Skip to Content
Merck
All Photos(1)

Documents

24381C

EX-CELL® CD Insect Cell Medium

Chemically defined, powder, suitable (for insect cell culture)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Quality Level

product line

EX-CELL®

form

dry powder

technique(s)

cell culture | insect: suitable

components

L-glutamine: yes

shipped in

ambient

storage temp.

2-8°C

General description

EX-CELL® CD Insect Cell Medium can be used as amplification and production medium for Spodoptera frugiperda (Sf) cells such as Sf21 and Sf9 cells and is compatible with Tni, C636 and S2 cells. The medium has been optimized to get excellent growth and productivity for the rhabdovirus-free Sf-RVN Insect Cell Line(Sf9). Together those products form the Sf-RVN Platform. The formulation is chemically defined, animal-component free and has been formulated with L-Glutamine.

Quantity

Formulated to contain 31.7 grams of powder per liter of medium.

Legal Information

EX-CELL is a registered trademark of Merck KGaA, Darmstadt, Germany
Sf-RVN is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

See how the Sf9 rhabdovirus-free cell line was developed and how we’ve developed a companion chemically-defined insect cell media for protein and viral expression.

To address scalability challenges of AAV manufacturing, we developed an HEK293 suspension cell line that can be used across many serotypes. Get the data in this article.

Minimize the complexities of cell and gene therapy production. Switch to upstream viral vector platforms to enhance scalability and simplify GMP manufacturing.

The current status and future manufacturing of AAV-based gene therapies is discussed in this podcast transcript, including how to streamline large-scale manufacturing.

See All

Related Content

A transfection-based solution to viral vector production using a suspension cell line, chemically defined medium, and a process with proven performance at scale.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service