Skip to Content
Merck
All Photos(1)

Key Documents

21236

Sigma-Aldrich

Calcium D-saccharate tetrahydrate

98.5-102% (KT)

Synonym(s):

D-Glucaric acid calcium salt, D-Saccharic acid calcium salt, Calcium D-glucarate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$191.00

$191.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$191.00

About This Item

Empirical Formula (Hill Notation):
C6H8CaO8 · 4H2O
CAS Number:
Molecular Weight:
320.26
Beilstein:
3821334
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

$191.00


Check Cart for Availability

Request a Bulk Order

biological source

synthetic

Assay

98.5-102% (KT)

form

powder

optical activity

[α]20/D +20.5±2°, c = 6% in 5 M HCl

color

white

storage temp.

room temp

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].O[C@H]1[C@H](O)[C@@H](O)C(=O)O[Ca]OC(=O)[C@H]1O

InChI

1S/C6H10O8.Ca.4H2O/c7-1(3(9)5(11)12)2(8)4(10)6(13)14;;;;;/h1-4,7-10H,(H,11,12)(H,13,14);;4*1H2/q;+2;;;;/p-2/t1-,2-,3-,4+;;;;;/m0...../s1

InChI key

ZQWFSIZRQANUDA-WQMSYZFBSA-L

Looking for similar products? Visit Product Comparison Guide

Application

D-Saccharate (D-glucaric acid) is used to study its potential chemopreventive properties and anti-oxidative properties at the level of peroxynitrite-induced damages.

Biochem/physiol Actions

Calcium glucarate is a constituent of many fruits (apples, citrus fruits) and cruciferous vegetables. Inhibits β-glucuronidase, a phase II enzyme that is elevated in some hormone-dependent cancers of the breast, prostate, and colon.

Other Notes

To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service