Skip to Content
Merck
All Photos(1)

Key Documents

17806

Sigma-Aldrich

Senecionine

≥95.0% (GC)

Synonym(s):

Aureine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H25NO5
CAS Number:
Molecular Weight:
335.39
Beilstein:
8162955
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

Assay

≥95.0% (GC)

optical activity

[α]25/D −55.1°, c = 0.034% in chloroform

mp

236 °C (lit.)

storage temp.

−20°C

SMILES string

[H][C@@]12[C@H]3CCN1CC=C2COC(=O)[C@](C)(O)[C@H](C)C\C(=C\C)C(=O)O3

InChI

1S/C18H25NO5/c1-4-12-9-11(2)18(3,22)17(21)23-10-13-5-7-19-8-6-14(15(13)19)24-16(12)20/h4-5,11,14-15,22H,6-10H2,1-3H3/b12-4-/t11-,14-,15-,18-/m1/s1

InChI key

HKODIGSRFALUTA-JTLQZVBZSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Senecionine has been used in a study to observe the gender-dependent difference of glutathione antioxidant system and its influence on hepatotoxic pyrrolizidine alkaloid isoline-induced liver injury. Senecionine has also been used in a study to observe a new metabolic pathway for pyrrolizidine alkaloids.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 1 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qingning Liang et al.
Toxicology, 280(1-2), 61-69 (2010-12-04)
Intracellular reduced glutathione (GSH) antioxidant system is crucial for counteracting oxidative stress-induced liver injury. The present study was designed to observe the gender-dependent difference of GSH antioxidant system and its influence on hepatotoxic pyrrolizidine alkaloid (HPA) isoline-induced liver injury. Lower
Changhong Wang et al.
Analytical and bioanalytical chemistry, 401(1), 275-287 (2011-05-17)
Pyrrolizidine alkaloids (PAs) are considered to be one of the most hepatotoxic groups of compounds of plant origin and are present in about 3% of the world's flowering plants. Most PAs represent a considerable health hazard to both livestock and
Fabiana Elias et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 49(9), 2313-2319 (2011-07-05)
This study is the first in the literature to focus attention on the possible immunotoxic effect of integerrimine N-oxide content in the butanolic residue (BR) of Senecio brasiliensis, a poisonous hepatotoxic plant that contains pyrrolizidine alkaloids (PAs). PAs have been
Pyrrolizidine alkaloids from Gynura divaricata.
E Roeder et al.
Planta medica, 62(4), 386-386 (1996-08-01)
V R Campesato et al.
Environmental and molecular mutagenesis, 29(1), 91-97 (1997-01-01)
Integerrimine (ITR), a pyrrolizidine alkaloid from Senecio brasiliensis, was tested for genotoxicity using the wing somatic mutation and recombination test (SMART) in Drosophila melanogaster. The compound was administered by chronic feeding (48 hours) of 3-day-old larvae. Two different crosses involving

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service