Skip to Content
Merck
All Photos(1)

Key Documents

17375

Sigma-Aldrich

Mag-Indo 1-AM

for fluorescence, ≥95% (HPLC)

Synonym(s):

Indo 1-AM analog

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H34N2O17
CAS Number:
Molecular Weight:
730.63
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

for fluorescence

Assay

≥95% (HPLC)

solubility

DMSO: soluble

fluorescence

λex 328 nm; λem 416 nm in 0.1 M Tris pH 8.0, esterase; Mg2+

storage temp.

2-8°C

SMILES string

CC(=O)OCOC(=O)COc1cc(ccc1N(CC(=O)OCOC(C)=O)CC(=O)OCOC(C)=O)-c2cc3ccc(cc3[nH]2)C(=O)OCOC(C)=O

InChI

1S/C47H51N3O22/c1-28-7-11-39(49(19-43(56)68-23-63-29(2)51)20-44(57)69-24-64-30(3)52)41(15-28)61-13-14-62-42-18-35(37-16-34-8-9-36(17-38(34)48-37)47(60)72-27-67-33(6)55)10-12-40(42)50(21-45(58)70-25-65-31(4)53)22-46(59)71-26-66-32(5)54/h7-12,15-18,48H,13-14,19-27H2,1-6H3

InChI key

CAWBRCOBJNWRLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Cell permeable fluorescent derivative of indicator for Mg2+

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Josué Villegas-Mendoza et al.
International journal of molecular sciences, 20(3) (2019-02-15)
In the ocean, the prokaryote respiration rates dominate the oxidation of organics, but the measurements may be biased due to pre-incubation size filtration and long incubation times. To overcome these difficulties, proxies for microbial respiration rates have been proposed, such
Philippe O Gannon et al.
Journal of immunology (Baltimore, Md. : 1950), 195(1), 356-366 (2015-05-24)
Experimental models demonstrated that therapeutic induction of CD8 T cell responses may offer protection against tumors or infectious diseases providing that T cells have sufficiently high TCR/CD8:pMHC avidity for efficient Ag recognition and consequently strong immune functions. However, comprehensive characterization
Michael Hebeisen et al.
Cancer research, 75(10), 1983-1991 (2015-03-27)
The avidity of the T-cell receptor (TCR) for antigenic peptides presented by the peptide-MHC (pMHC) on cells is a key parameter for cell-mediated immunity. Yet a fundamental feature of most tumor antigen-specific CD8(+) T cells is that this avidity is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service