Skip to Content
Merck
All Photos(1)

Key Documents

14581C

SAFC

EX-CELL® MDCK Serum-Free Medium for MDCK Cells

Animal-protein free, without L-glutamine, liquid, sterile-filtered, suitable for cell culture

Sign Into View Organizational & Contract Pricing

Select a Size

1000 ML
$218.00

$218.00


Check Cart for Availability


Select a Size

Change View
1000 ML
$218.00

About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

$218.00


Check Cart for Availability

description

for research or for further manufacturing use

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

components

glucose: 6.0 g/L
NaHCO3: 1.8 g/L
HEPES: no
L-glutamine: no

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

EX-CELL MDCK is an animal-protein free, serum-free medium developed for the long-term growth of Madin Darby Canine Kidney (MDCK) and related cells. The cells, in an attachment culture, can be subcultured directly into EX-CELL MDCK from serum-supplemented media without adaptation. Cell densities and doubling times achieved under serum-free conditions are comparable to those achieved in a serum-supplemented culture.

Legal Information

EX-CELL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service