1446B
BIOEAZE™ Ethyl Vinyl Acetate (EVA) Bags
volume 350 mL, designed for use with Terumo Aseptic Connecting Device (ACD) Applications
About This Item
sterility
sterile; γ-irradiated
packaging
pkg of 10 bags
volume
350 mL
Looking for similar products? Visit Product Comparison Guide
General description
Port P1: Swabable Luer Lock
Port P2: 1 x 1.5 ft. length of 1/8" x 1/4" clear 082 C-Flex® tubing with pinch clamp and plug
Port P3: 1 Spike Port
Features and Benefits
BIOEAZE Ethyl Vinyl Acetate (EVA) bags are constructed from a single-web, medical grade laminate of four different films, with a fluid contact surface of EVA, with two layers of low-density polyethylene (LDPE) enveloping a layer of ethyl vinyl alcohol (EVOH).
Packaging
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service