Skip to Content
Merck
All Photos(1)

Key Documents

14448

Sigma-Aldrich

2-(4-Biphenylyl)-6-phenylbenzoxazole

BioReagent, suitable for scintillation, ≥98.0% (TLC)

Synonym(s):

PBBO

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$90.40

$90.40


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
1 G
$90.40

About This Item

Empirical Formula (Hill Notation):
C25H17NO
CAS Number:
Molecular Weight:
347.41
Beilstein:
1222425
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

$90.40


Estimated to ship on31 May 2025


Request a Bulk Order

product line

BioReagent

Assay

≥98.0% (TLC)

form

solid

mp

198-199 °C (lit.)

fluorescence

λem 390 nm in acetonitrile: methanol
λem 425 nm in H2O

suitability

suitable for scintillation

SMILES string

c1ccc(cc1)-c2ccc(cc2)-c3nc4ccc(cc4o3)-c5ccccc5

InChI

1S/C25H17NO/c1-3-7-18(8-4-1)20-11-13-21(14-12-20)25-26-23-16-15-22(17-24(23)27-25)19-9-5-2-6-10-19/h1-17H

InChI key

WISWLZYHZLVSMO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Utilized as a scintillator reagent[1][2][3].

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evaluation of a photoelectron-rejecting alpha liquid-scintillation (PERALS) spectrometer for the measurement of alpha-emitting radionuclides
Cadieux, J. R.
Nuclear Instruments & Methods in Physics Research. Section A, Accelerators, Spectrometers, Detectors and Associated Equipment, 299, 119-119 (1990)
Some rules to improve the energy resolution in alpha liquid scintillation with beta rejection
Aupiais, J., Aubert, C., and Dacheux, N.
Radiochim. Acta, 91, 63-63 (2003)
Genetics of resistance of pyrethroid-selected houseflies, Musca domestica L.
Farnham, A. W.
Pestic. Sci., 4, 513-513 (1973)
Dimitrios Kioroglou et al.
Foods (Basel, Switzerland), 9(10) (2020-10-03)
Wine origin and ageing are two factors related to wine quality which in turn is associated to wine metabolome. Currently, new metabolomic techniques and proper statistics procedures allow accurate profiling of wine metabolome. Thus, the main goal was to evaluate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service