141-500A
EpiVita Growth Medium for Adult Keratinocytes, Serum-Free
Select a Size
$242.00
Select a Size
About This Item
$242.00
form
liquid
Quality Level
shelf life
6 mo.
manufacturer/tradename
Cell Applications, Inc
storage condition
protect from light
shipped in
wet ice
storage temp.
2-8°C
General description
Quality
Preparation Note
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service