Key Documents
1336R
Millipore
BIOEAZE Rigging Sets
About This Item
UNSPSC Code:
12352200
description
Designed for use with Quick Connect Applications with Male and Female Quick Connects with Caps, Double Polybagged
sterility
sterile; γ-irradiated
General description
1 x 10 ft. length of Size 15 Opaque 001 C-Flex® Tubing and 1/4" Quick Connects with Caps
Packaging
Double Polybagged
Legal Information
BIOEAZE is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)