Skip to Content
Merck
All Photos(1)

Key Documents

12121201

Sigma-Aldrich

B9 Cell Line mouse

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Synonym(s):

B9 Cells

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
$1,560.00

$1,560.00


Check Cart for Availability


Select a Size

Change View
1 VIAL
$1,560.00

About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

$1,560.00


Check Cart for Availability

biological source

mouse spleen

description

Mouse B cell hybridoma, IL-6 assay

packaging

tube of 5 μg 12121201-DNA-5UG
pkg of vial of cells 12121201-1VL

growth mode

Suspension

karyotype

Not specified

morphology

Lymphoblastoid

technique(s)

cell culture | mammalian: suitable

Cell Line Origin

Mouse B cell hybridoma, IL-6 assay

Cell Line Description

B9 is a mouse B cell hybridoma line that requires Interleukin 6 (IL-6) for survival and proliferation. IL-6 is a pluripotent cytokine with multiple effects on many different cell types. It is produced by a variety of cells in response to various stimuli. The B9 hybridoma allows for a simple sensitive assay for human, rat, rabbit, pig and mouse IL-6 based on the IL-6 dependent proliferation of this murine B cell line. Details of the B9 hybridoma growth factor assay can be found in Current protocols in Immunology 1996 Cytokines and Their Cellular receptors 6.6.1-6.6.5 contributed by Nordan RP, Richards CD and Gauldie J. John Wiley & Sons, Inc.

Packaging

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Culture Medium

IDMEM or RPMI 1640 + 2mM Glutamine + 50μM 2-Mercaptoethanol (2ME) + 50pg/ml recombinant human IL-6 (Sigma SRP3096, diluted and stored in PBS containing 1% BSA) + 5% FBS. IL-6 needs to be added fresh at each media change.

Subculture Routine

For B9 cell maintenance cells should be seeded at 0.5 x 105 cells/ml in B9 culture medium. The culture will be in log phase growth between days 3-5. When the culture reaches 1 x 106 cells/ml (4-5 days) the culture must be sub cultured as above again seeding at 0.5 x 105 cells/ml.; 5% CO2; 37°C. It maybe necessary to determine the activity of the IL-6 or IL-6 containing supernatant first. The lowest concentration at which maximal growth can be obtained should be used. A fresh vial of cells is required when the background response from control cultures begins to rise. It is important that the culture conditions are adhered to avoid the B9 cells becoming IL-6 independent.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service