Skip to Content
Merck
All Photos(1)

Documents

11640

Sigma-Aldrich

Azophloxine

for microscopy (Hist.)

Synonym(s):

Acid Red 1, Food Red 10

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H13N3Na2O8S2
CAS Number:
Molecular Weight:
509.42
Colour Index Number:
18050
Beilstein:
4121767
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

for microscopy (Hist.)

form

powder

solubility

methanol: water (1:1): 0.5 g/10 mL, red

εmax

≥300 at 527-537 nm in water

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].CC(=O)Nc1cc(cc2cc(c(\N=N\c3ccccc3)c(O)c12)S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C18H15N3O8S2.2Na/c1-10(22)19-14-9-13(30(24,25)26)7-11-8-15(31(27,28)29)17(18(23)16(11)14)21-20-12-5-3-2-4-6-12;;/h2-9,23H,1H3,(H,19,22)(H,24,25,26)(H,27,28,29);;/q;2*+1/p-2/b21-20+;;

InChI key

WXLFIFHRGFOVCD-SERMZQFOSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Azophloxine, also known as acid red 1 (AR 1), is a member of synthetic red azo dye family. Azo dye is an organic dye with good photo-thermal stability, dissolvability and easy preparation feature. In addition, it also has noticeable third order nonlinearity and good optical power limiting properties. It has been used as a real acid counterstain in light and fluorescence microscopy. Azophloxine also facilitates the determination of proteins and peptides.

Application

Azophloxine has been used as a dye in Von Kossa staining and nonlinear optical studies. It is also used in photocatalytic tests.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Intrinsic differentiation potential of adolescent human tendon tissue: an in-vitro cell differentiation study.
de Mos M
BMC Musculoskelet. Disord., 8, 16-16 (2007)
Photosensitization of CuI - the role of visible light induced CuI? CuII transition in photocatalytic degradation of organic pollutants and inactivation of microorganisms.
Wojtyla S
Photochemical & Photobiological Sciences : Official Journal of the European Photochemistry Association and the European Society for Photobiology, 16(7), 1079-1087 (2017)
Investigation of Optical limiting properties of Azophloxine
dye using nanosecond Z- scan technique
Parvin M
International Journal of ChemTech Research, 6, 2493-2498 (2014)
A Voltammetric Study on the Aqueous Electrochemistry of
Acid Red 1 (Azophloxine)
Bicer E and Arat C
Croatica Chemica Acta. Arhiv Za Kemiju, 82(3), 583?593-583?593 (2009)
Histochemical detection of myoepithelial cells in pleomorphic adenoma with azophloxine and amidoblack.
A G Triantafyllou
Journal of oral medicine, 41(3), 184-188 (1986-07-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service