Skip to Content
Merck
All Photos(1)

Key Documents

08075

Sigma-Aldrich

Potassium β-hydroxypyruvate

≥95.0% (NT)

Synonym(s):

β-Hydroxypyruvic acid potassium salt, 3-Hydroxy-2-oxopropionic acid potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2COCOOK
CAS Number:
Molecular Weight:
142.15
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Assay

≥95.0% (NT)

form

powder

SMILES string

[K+].OCC(=O)C([O-])=O

InChI

1S/C3H4O4.K/c4-1-2(5)3(6)7;/h4H,1H2,(H,6,7);/q;+1/p-1

InChI key

GQHLMYFYRZSQRJ-UHFFFAOYSA-M

Biochem/physiol Actions

Hydroxypyruvate is a metabolite involved in the pathway of carbon in photorespiration.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Edward G Hibbert et al.
Journal of biotechnology, 131(4), 425-432 (2007-09-11)
We have used active-site targeted directed evolution by saturation mutagenesis to improve the activity of E. coli transketolase towards non-phosphorylated substrates. Residues were selected for each set based on either structural proximity to substrate, or on phylogenetic variation. Each library
C V Givan et al.
Plant physiology, 100(2), 552-556 (1992-10-01)
Glyoxylate and hydroxypyruvate are metabolites involved in the pathway of carbon in photorespiration. The chief glyoxylate-reducing enzyme in leaves is now known to be a cytosolic glyoxylate reductase that uses NADPH as the preferred cofactor but can also use NADH.
Stefan Timm et al.
Plant physiology, 155(2), 694-705 (2011-01-06)
Hydroxypyruvate (HP) is an intermediate of the photorespiratory pathway that originates in the oxygenase activity of the key enzyme of photosynthetic CO(2) assimilation, Rubisco. In course of this high-throughput pathway, a peroxisomal transamination reaction converts serine to HP, most of
Kai Yang et al.
Chemico-biological interactions, 191(1-3), 315-321 (2011-03-08)
Previously, we showed that dietary fructose or its carbonyl metabolites, glyceraldehyde and glycolaldehyde, could be oxidized by inflammatory reactive oxygen species (ROS), products of immune cells, to form highly toxic and genotoxic products, such as glyoxal. Glycolaldehyde-caused hepatocyte protein carbonylation
D Pastore et al.
Plant & cell physiology, 42(12), 1373-1382 (2002-01-05)
In order to gain a first insight into the alternative oxidase (AO) function in durum wheat mitochondria (DWM), we investigated some activation pathways of this enzyme in DWM purified from both etiolated shoots and green leaves. AO was activated when

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service