Skip to Content
Merck
All Photos(1)

Documents

07668

Sigma-Aldrich

Echinacoside

98% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H46O20
CAS Number:
Molecular Weight:
786.73
Beilstein:
4778612
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.32

Assay

98% (HPLC)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@H](OCCc3ccc(O)c(O)c3)O[C@H](CO[C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O)[C@H]2OC(=O)\C=C\c5ccc(O)c(O)c5)[C@H](O)[C@H](O)[C@H]1O

InChI

1S/C35H46O20/c1-14-24(42)26(44)29(47)35(51-14)55-32-30(48)34(49-9-8-16-3-6-18(38)20(40)11-16)53-22(13-50-33-28(46)27(45)25(43)21(12-36)52-33)31(32)54-23(41)7-4-15-2-5-17(37)19(39)10-15/h2-7,10-11,14,21-22,24-40,42-48H,8-9,12-13H2,1H3/b7-4+/t14-,21+,22+,24-,25+,26+,27-,28+,29+,30+,31+,32+,33+,34+,35-/m0/s1

InChI key

FSBUXLDOLNLABB-ISAKITKMSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Echinacoside (ECH) is a natural secondary metabolite, a phenylethanoid glycoside (PhG) found in the roots and stems of many plants. It is a water-soluble compound. Echinacoside was first isolated from the roots of Echinacea angustifolia (coneflower). It can also be sourced from other medicinal plants such as Echinacea purpurea (purple coneflower) and Cistanche salsa.

Biochem/physiol Actions

Caffeic acid glycoside found in herbal medicine. Potential therapeutic for Parkinson′s disease.
Echinacoside elicits neuroprotective properties against neurological disorders and on tumor necrosis factor-alpha (TNFα)-induced apoptosis in human neuroblastoma (SH-SY5Y) cells. It also displays antioxidant and free radical scavenging properties. ECH exerts hepatoprotective and anti-cancer properties.
Echinacoside is a secondary metabolite found in plants, specifically in the roots of Echinacea angustifolia (cone flower) , E. purpurea (purple coneflower) and Cistanche salsa. It has been shown to possess anti-oxidant and free radical scavenging properties, to have neuroprotective effects and to prevent liver injuries. It has also been found to be antibiotic and anti-inflammatory. Echinacoside was shown to promote proliferation of intestinal epithelial cells by increasing TGF-β1 expression.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C E Bomberger et al.
Thymus, 19(2), 111-115 (1992-03-01)
Murine bone marrow cells were examined in an in vitro assay to determine whether stress modulates the migration of prethymic stem cells to thymus supernatant. Adult CBA/J mice were either restraint or sound stressed for two hours daily for five
Monika E Czerwińska et al.
Phytochemical analysis : PCA, 26(4), 253-260 (2015-02-20)
The genus Ligustrum (Oleaceae) is distributed in Europe and Asia (south China and Korea), where it is used to prevent hypertension, sore throats, inflammation and diabetes. The main groups of compounds in extracts of Ligustrum vulgare are biologically active secoiridoids
N Shiotani et al.
Veterinary parasitology, 41(1-2), 17-22 (1992-02-01)
The distribution of oocysts, sporocysts and sporozoites of Eimeria tenella and Eimeria maxima in the digestive tract of chicken and in excreta was investigated. At 1 h after the oral inoculation of E. tenella oocysts, the number of sporocysts in
Rong Kuang et al.
Planta medica, 75(14), 1499-1504 (2009-06-24)
We have investigated the protective effects of echinacoside (ECH), one of the phenylethanoid glycosides, on H(2)O(2)-induced cytotoxicity in the rat pheochromocytoma cell line (PC12 cells). Our data show that application of ECH to H(2)O(2)-injured PC12 cells (HIPCs) increased cell viability
Parida Yamada et al.
Planta medica, 76(14), 1512-1518 (2010-04-01)
The immediate-type allergic reaction is involved in many allergic diseases such as asthma, allergic rhinitis, and sinusitis. In this study, we investigated the effect of acteoside extracted from CISTANCHE TUBULOSA (Schrenk) R. Wight on the basophilic cell-mediated allergic reaction. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service