Skip to Content
Merck
All Photos(1)

Key Documents

04449

Sigma-Aldrich

O-(Octadecylphosphoryl)choline

≥98% (TLC)

Synonym(s):

Octadecylphosphocholine, Phosphocholine octadecyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H50NO4P
CAS Number:
Molecular Weight:
435.62
Beilstein:
3694312
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

Assay

≥98% (TLC)

lipid type

phospholipids

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCCCCOP([O-])(=O)OCC[N+](C)(C)C

InChI

1S/C23H50NO4P/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-22-27-29(25,26)28-23-21-24(2,3)4/h5-23H2,1-4H3

InChI key

ZBNJXSZNWZUYCI-UHFFFAOYSA-N

Application

Antitumor phospholipid, induces apoptosis in three human leukemic cell lines.

Biochem/physiol Actions

C18-phosphocholine was the most potent alkylphosphocholine tested in inhibiting phosphatidylcholine biosynthesis. The effect is mediated by interrupting the translocation of the rate-limiting enzyme, CTP:phosphocholine cytidylyltransferase, to membranes, where it is active.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Zeisig et al.
Anti-cancer drugs, 2(4), 411-417 (1991-08-01)
Hexadecylphosphocholine (HPC) and its analogs with a longer alkyl chain (C18 and C20) were examined for antineoplastic activity in the murine tumor models P388 leukemia, B 16 melanoma, the mammary carcinoma C3H and Ca 755, and the human MT-1 mammary
V Brochez et al.
Lipids, 34(5), 511-516 (1999-06-24)
The determination of cellular content of octadecylphosphocholine (D-19391) and hexadecylphosphocholine (HePC, D-18506), two anticancer agents of the alkylphosphocholine group, using capillary gas chromatography is described. The compounds' cytotoxicity was first determined by the MTT [3-(4,5-dimethyl-2-thiazolyl)-2,5-diphenyltetrazolium] assay, being indicative for the
James McBride et al.
Journal of clinical microbiology, 43(2), 629-634 (2005-02-08)
We have developed and optimized a 96-well microtiter plate assay, based on the reduction of alamarBlue, to assess the efficacies of much needed new antimicrobials against Acanthamoeba species. This assay has been optimized for determination of drug efficacy against two
E Posse de Chaves et al.
The Biochemical journal, 312 ( Pt 2), 411-417 (1995-12-01)
At least 50% of the major axonal membrane lipid, phosphatidylcholine, of rat sympathetic neurons is synthesized in situ in axons [Posse de Chaves, Vance, Campenot and Vance (1995) J. Cell Biol. 128, 913-918]. In the same study we reported that
N Dittrich et al.
Journal of enzyme inhibition, 11(1), 67-75 (1996-08-01)
Alkylphosphate esters were shown to be potent inhibitors of phospholipase D. Using phosphatidyl choline/sodium dodecylsulfate (2:1) as substrate, IC50 values were determined for alkylphosphocholines of different chain length (C10-C18) and for various octadecylphosphate esters with different polar head groups. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service