Skip to Content
Merck
All Photos(1)

Key Documents

04394

Sigma-Aldrich

Peroxidase, Nitrospira sp., recombinant

≥10.0 U/g

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$139.00

$139.00


Estimated to ship on28 April 2025



Select a Size

Change View
10 MG
$139.00

About This Item

CAS Number:
EC Number:
UNSPSC Code:
12352204
NACRES:
NA.54

$139.00


Estimated to ship on28 April 2025


biological source

bacterial (Nitrospira spp.)

Quality Level

specific activity

≥10.0 U/g

storage temp.

−20°C

SMILES string

[O+H2]O[O-]

InChI

1S/H2O3/c1-3-2/h1-2H

InChI key

JSPLKZUTYZBBKA-UHFFFAOYSA-N

Unit Definition

1 Unit corresponds to the amount which oxidizes 1 μmol of ABTS per minute at 25°C and pH 5.0

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service