Skip to Content
Merck
All Photos(1)

Key Documents

03714

Sigma-Aldrich

Blue Dextran

average mol wt 20,000

Synonym(s):

Blue Dextran 20, Dextran blue from Leuconostoc mesenteroides

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$377.00

$377.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
1 G
$377.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.32

$377.00


In StockDetails


Request a Bulk Order

biological source

synthetic

form

powder

mol wt

average mol wt 20,000

loss

≤10% loss on drying

capacity

0.01-0.08 mmol/g loading

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lorena Moreno-Vilet et al.
Food chemistry, 237, 833-840 (2017-08-03)
Agave fructans are increasingly important in food industry and nutrition sciences as a potential ingredient of functional food, thus practical analysis tools to characterize them are needed. In view of the importance of the molecular weight on the functional properties

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service