Skip to Content
Merck
All Photos(1)

Key Documents

S0882

Sigma-Aldrich

Sodium selenate

≥95% (elemental analysis)

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$122.00
100 G
$381.00

$122.00


Available to ship on09 April 2025Details


Request a Bulk Order

Select a Size

Change View
10 G
$122.00
100 G
$381.00

About This Item

Linear Formula:
Na2SeO4
CAS Number:
Molecular Weight:
188.94
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55
grade:
for analytical purposes
form:
powder or crystals

$122.00


Available to ship on09 April 2025Details


Request a Bulk Order

grade

for analytical purposes

Quality Level

Assay

≥95% (elemental analysis)

form

powder or crystals

pH

5.5-7.5 (25 °C, 18.9 g/L)

SMILES string

[Na+].[Na+].[O-][Se]([O-])(=O)=O

InChI

1S/2Na.H2O4Se/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

MHQOTKLEMKRJIR-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Selenium nanoparticles alleviate ischemia reperfusion injury-induced acute kidney injury by modulating GPx-1/NLRP3/Caspase-1 pathway.: This study demonstrates that selenium nanoparticles, including sodium selenate, mitigate acute kidney injury caused by ischemia reperfusion. The nanoparticles modulate key pathways, offering potential therapeutic applications in renal injury treatment (Wang et al., 2022).

  • Phosphoproteomic profiling of selenate-treated Alzheimer′s disease model cells.: This research provides a detailed phosphoproteomic profile of Alzheimer′s disease model cells treated with sodium selenate, uncovering its effects on protein phosphorylation and implications for neuroprotection (Chen et al., 2014).

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2 - STOT RE 2

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adam J Matich et al.
Phytochemistry, 75, 140-152 (2011-12-27)
Glucosinolates are sulphur-containing glycosides found in many Brassica spp. that are important because their aglycone hydrolysis products protect the plant from herbivores and exhibit anti-cancer properties in humans. Recently, synthetically produced selenium analogues have been shown to be more effective
M Djanaguiraman et al.
Plant physiology and biochemistry : PPB, 48(12), 999-1007 (2010-10-19)
Oxidative stress is commonly induced when plants are grown under high temperature (HT) stress conditions. Selenium often acts as an antioxidant in plants; however, its role under HT-induced oxidative stress is not definite. We hypothesize that selenium application can partly
Maria Łabanowska et al.
Journal of plant physiology, 169(3), 275-284 (2011-12-14)
Biochemical analyses of antioxidant content were compared with measurements of fluorescence and electron paramagnetic resonance (EPR) to examine the alteration of radicals in wheat seedlings exposed to 2 days of selenium stress. Two genotypes of Polish and one of Finnish
W C Stewart et al.
Journal of animal science, 90(2), 577-584 (2011-10-04)
Adequate Se transfer from ewes to lambs is important to prevent Se-deficiency diseases. To evaluate how different chemical forms of Se administered at comparative dosages to mature ewes affect Se status of their lambs, 240 ewes were divided into 8
Beatrice Pezzarossa et al.
Journal of the science of food and agriculture, 92(4), 781-786 (2011-09-29)
We investigated the effects of the foliar and fruit application of sodium selenate on selenium (Se) accumulation, fruit growth and ripening in peach and pear. Trials were conducted in two growing seasons. In 2008 selenate was applied at a rate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service