Skip to Content
Merck
All Photos(1)

Key Documents

908924

Sigma-Aldrich

Tin(II) chloride

free-flowing, Redi-Dri, reagent grade, 98%

Synonym(s):

Stannous chloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$136.00
500 G
$378.00

$136.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$136.00
500 G
$378.00

About This Item

Linear Formula:
SnCl2
CAS Number:
Molecular Weight:
189.62
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.21
grade:
anhydrous
reagent grade
form:
crystals

$136.00


Check Cart for Availability

Request a Bulk Order

grade

anhydrous
reagent grade

product line

Redi-Dri

Assay

98%

form

crystals

quality

free-flowing

bp

652 °C (lit.)

mp

246 °C (lit.)

SMILES string

Cl[SnH2]Cl

InChI

1S/2ClH.Sn/h2*1H;/q;;+2/p-2

InChI key

AXZWODMDQAVCJE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

SnCl2 dihydrate reacts with acetic anhydride to yield anhydrous SnCl2. SnCl2 exhibits reducing properties in acidic media. SnCl2 shows superior catalytic action in its molten state. It is a promising raw material for the chemical vapour deposition (CVD) of semiconducting layers. SnCl2 participates in the reduction of aromatic nitro compounds, nitriles, cyanosilyl ethers and organic azides.

Application

Redi-Dri is an innovative packaging product line that helps eliminate clumping material by adding extra packaging process steps without any chemical additives, anti-clumping agents, or hydrophobic compounds. Redi-Drioffers a more convenient and safer way to handle the products.
Tin(II) chloride (SnCl2) has been used as a starting material to evaluate the thermal behavior of SnCl2 on heating at the rate of 5°Cmin-1 in air. The product undergoes phase transformation on heating at rate of 5°C min-1 in air, it involves an incomplete evaporation followed by oxidation forming SnO2 nanoparticles. SnCl2 has also been used for the fabrication of Sn-containing carbon nano-structures.
Tin(II) chloride may be used:
  • To catalyze the addition of diazo sulfones, diazo phosphine oxides and diazo phosphonates to aldehydes to form β-keto sulfones, β-keto phosphine oxides and β-keto phosphonates, respectively.
  • Along with trityl chloride, to catalyze the aldol reaction of silyl enol ethers with acetals or aldehydes and the Michael reaction of silyl enol ethers with α,β-unsaturated ketones.
  • As a promoter in the allylic amination of allylic alcohols with amines in the presence of palladium catalyst.

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 2 Oral - STOT SE 3

Target Organs

Cardio-vascular system, Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service