Skip to Content
Merck
All Photos(1)

Key Documents

77310

Sigma-Aldrich

Periodic acid

puriss. p.a., ACS reagent, for oxidimetric titration, crystallized, ≥99.0% (RT)

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$126.00
100 G
$480.00

$126.00


Estimated to ship on02 April 2025


Request a Bulk Order

Select a Size

Change View
25 G
$126.00
100 G
$480.00

About This Item

Empirical Formula (Hill Notation):
H5IO6
CAS Number:
Molecular Weight:
227.94
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21

$126.00


Estimated to ship on02 April 2025


Request a Bulk Order

grade

ACS reagent
for oxidimetric titration
puriss. p.a.

Quality Level

Assay

≥99.0% (RT)

form

powder or crystals

quality

crystallized

ign. residue

≤0.01% (as SO4)

mp

122 °C (lit.)

solubility

water: soluble

anion traces

bromide, bromate, chloride, chlorate (as Cl-): ≤100 mg/kg
iodide (I-): ≤10 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

Ag: ≤5 mg/kg
Al: ≤5 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤30 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤10 mg/kg
Sr: ≤5 mg/kg
Tl: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

OI(O)(O)(O)(O)=O

InChI

1S/H5IO6/c2-1(3,4,5,6)7/h(H5,2,3,4,5,6,7)

InChI key

TWLXDPFBEPBAQB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Periodic acid (H5IO6) is a colorless crystalline solid. It is widely employed as an oxidant and is weakly acidic in nature. It can be synthesized from iodic(V) acid.[1]

Application

Periodic acid may be used in the synthesis of diiodine heptoxide (I2O7).[2] It may be used in the Periodic acid Schiff staining procedures.[3]

Other Notes

for the histochemical detection of polysaccharides

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Ox. Sol. 1 - Skin Corr. 1B - STOT RE 1 Oral

Target Organs

Thyroid

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monica A Mainigi et al.
Biology of reproduction, 90(2), 26-26 (2013-12-20)
Assisted reproductive technologies (ART) have been associated with several adverse perinatal outcomes involving placentation and fetal growth. It is critical to examine each intervention individually in order to assess its relationship to the described adverse perinatal outcomes. One intervention ubiquitously
Eagleson M.
Concise Encyclopedia Chemistry, 791-791 (1994)
Eagleson M.
Concise Encyclopedia Chemistry, 543-543 (1994)
Murad Alturkustani et al.
Journal of neuropathology and experimental neurology, 74(3), 233-240 (2015-02-11)
The pathologic features of adult-onset leukoencephalopathy/leukodystrophy with axonal spheroids (ALAS) are variable, and this has led to different hypotheses as to whether primarily demyelination or axonopathy may underlie this disorder. Typical ALAS pathology is rarely accompanied by focal multiple sclerosis
Francesca Polverino et al.
The American journal of pathology, 185(3), 741-755 (2014-12-30)
Small animal models of chronic obstructive pulmonary disease (COPD) have several limitations for identifying new therapeutic targets and biomarkers for human COPD. These include a pulmonary anatomy that differs from humans, the limited airway pathologies and lymphoid aggregates that develop

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service