Skip to Content
Merck
All Photos(1)

Key Documents

77240

Sigma-Aldrich

Peracetic acid solution

purum, ~39% in acetic acid (RT)

Synonym(s):

Peroxyacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO3H
CAS Number:
Molecular Weight:
76.05
Beilstein:
1098464
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21

grade

purum

form

liquid

shelf life

limited shelf life, expiry date on the label

reaction suitability

reagent type: oxidant

concentration

38.5-41.5% (iodometric, redox titration)
~39% in acetic acid (RT)

impurities

≤6% H2O2
~45% acetic acid

refractive index

n20/D 1.391

density

1.15 g/mL at 20 °C

storage temp.

2-8°C

SMILES string

CC(=O)OO

InChI

1S/C2H4O3/c1-2(3)5-4/h4H,1H3

InChI key

KFSLWBXXFJQRDL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The product is a 39% solution of peracetic acid in acetic acid. Peracetic acid is widely employed for the disinfection of sewage since it exhibits anti-bacterial, antifungal and sporicidal characteristics.[1]

Application

Peracetic acid may be used in the synthesis of oxiranes (epoxides) via Prilezhaev reaction.[2]

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Flam. Liq. 3 - Org. Perox. D - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 2

Flash Point(F)

132.8 °F - closed cup

Flash Point(C)

56 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eagleson M.
Concise Encyclopedia Chemistry, 743-743 (1994)
Mehmet Kitis
Environment international, 30(1), 47-55 (2003-12-11)
Peracetic acid is a strong disinfectant with a wide spectrum of antimicrobial activity. Due to its bactericidal, virucidal, fungicidal, and sporicidal effectiveness as demonstrated in various industries, the use of peracetic acid as a disinfectant for wastewater effluents has been
Alberto Baggioli et al.
Physical chemistry chemical physics : PCCP, 15(4), 1130-1140 (2012-12-12)
We examine several computational strategies for the prediction of the (17)O-NMR shielding constants for a selection of organic acids and peracids in aqueous solution. In particular, we consider water (the solvent and reference for the chemical shifts), hydrogen peroxide, acetic
D L Straus et al.
Journal of fish diseases, 35(7), 505-511 (2012-06-08)
Peracetic acid (PAA) is a relatively new compound suggested for use to treat pathogens in aquaculture. It is approved for use in Europe, but not in the United States. This study determined the effectiveness of PAA for fungus control on
Greetje A A Castelijn et al.
Veterinary microbiology, 161(3-4), 305-314 (2012-08-22)
Cross-contamination due to Salmonella on the surface of processing equipment greatly contributes to contamination of pork products. Therefore, a clear understanding of surface and survival behaviour of relevant Salmonella serovars in pork processing environments is needed to develop better strategies

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service