Skip to Content
Merck
All Photos(1)

Documents

745588

Sigma-Aldrich

Ethyl acetate/Ethanol 3:1 (v/v) solution

greener alternative

(Ethyl acetate solution with 26.2% v/v SDA 35A), suitable for HPLC

Synonym(s):

Ethyl acetate solution, Ethyl acetate solution, 3:1 (v:v) ethyl acetate: 200 proof ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
3 EtOAc/EtOH
UNSPSC Code:
12191502
NACRES:
NA.21

Quality Level

form

liquid

greener alternative product characteristics

Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

HPLC: suitable

impurities

≤0.01% (water)

density

0.868 g/mL at 25 °C

format

mixture

greener alternative category

InChI key

XEKOWRVHYACXOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This solution of Ethyl acetate/Ethanol is a greener alternative to dichloromethane for chromatographic purification and thus has been enhanced for "Safer Solvents and Auxiliaries". Click here for more information.

Application

Greener alternative to chlorinated solvents dichloromethane in chromatography.

Features and Benefits

Greener alternative for DCM

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

23.0 °F

Flash Point(C)

-5 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A convenient guide to help select replacement solvents for dichloromethane in chromatography
Taygerly, J. P.;
Green Chemistry, 14, 3020-3020 (2012)
Sustainable Practices in Medicinal Chemistry: Current State and Future Direction
Bryan, M. C.;
Journal of Medicinal Chemistry null

Related Content

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service