Skip to Content
Merck
All Photos(3)

Key Documents

688363

Sigma-Aldrich

Molecular sieves, 4 Å

powder, activated, −325 mesh particle size

Synonym(s):

SYLOSIV® A4, Sodium Alumino-Silicate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$53.00
500 G
$131.00
1 KG
$217.00
2.5 KG
$462.00

$53.00


Estimated to ship on30 March 2025


Request a Bulk Order

Select a Size

Change View
25 G
$53.00
500 G
$131.00
1 KG
$217.00
2.5 KG
$462.00

About This Item

Linear Formula:
Na12[(AlO2)12(SiO2)12] · xH2O
CAS Number:
MDL number:
UNSPSC Code:
23201100
eCl@ss:
32110204
NACRES:
NA.21

$53.00


Estimated to ship on30 March 2025


Request a Bulk Order

Quality Level

form

powder

particle size

−325 mesh

pore size

4 Å

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Molecular sieves are made up of three dimensional interconnecting networks of silica and alumina tetrahedral. These crystalline metal aluminosilicates have uniform cavities to selectively adsorb specific size molecules. A 4A molecular sieve has an effective pore size of 4Å.

Application

Molecules with an effective diameter of 4 Å, including H2O, H2S, CO2, SO2, C2H4 and C3H6 are adsorbed.

Legal Information

SYLOSIV is a registered trademark of W.R. Grace & Co.-Conn.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–5 of 5 Questions  
  1. What is the CEC for this zeolite? specifically for Ca2+ and Mg2+?

    1 answer
    1. The cation exchange capacity (CEC) of 4A molecular sieves is generally low, as these materials are primarily designed for adsorption and separation processes rather than for ion exchange. Typically, the CEC for 4A molecular sieves is around 0.1 to 0.5 meq/g. This capacity may vary depending on the specific formulation and conditions of the molecular sieve and in this case, it is not a tested parameter.

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  4. Hi, I am planning to buy Molecular sieves, 4 Å, product number 688363. As the product information says that this one is activated. Does this mean I can use it without any treatment to remove my solvent water?

    1 answer
    1. Yes, this item is a ready-to-use product. No treatment is required before use.

      Helpful?

  5. what is the effective protocol to use molecular sieve and recycle

    1 answer
    1. Please see the link below for general information, including regeneration, for molecular sieves:
      https://www.sigmaaldrich.com/technical-documents/technical-article/chemistry-and-synthesis/reaction-design-and-optimization/molecular-sieves

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service