Skip to Content
Merck
All Photos(1)

Key Documents

60832

Sigma-Aldrich

Cobalt(II) nitrate hexahydrate

puriss. p.a., ACS reagent, Ni ≤0.001%, ≥99.0% (KT)

Synonym(s):

Cobaltous nitrate, Cobaltous nitrate hexahydrate, Nitric acid, cobalt(II) salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Co(NO3)2 · 6H2O
CAS Number:
Molecular Weight:
291.03
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NB.24
form:
powder or crystals

grade

ACS reagent
puriss. p.a.

Assay

≥99.0% (KT)

form

powder or crystals

reaction suitability

reagent type: catalyst
core: cobalt

pH

4 (20 °C, 100 g/L)

mp

55 °C (lit.)

anion traces

chloride (Cl-): ≤10 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤10 mg/kg
Cd: ≤10 mg/kg
Cr: ≤50 mg/kg
Cu: ≤10 mg/kg
Fe: ≤10 mg/kg
K: ≤100 mg/kg
Mg: ≤10 mg/kg
Mn: ≤20 mg/kg
NH4+: ≤500 mg/kg
Na: ≤100 mg/kg
Ni: ≤10 mg/kg
Pb: ≤10 mg/kg
Zn: ≤50 mg/kg

SMILES string

O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Co.2NO3.6H2O/c;2*2-1(3)4;;;;;;/h;;;6*1H2/q+2;2*-1;;;;;;

InChI key

QGUAJWGNOXCYJF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cobalt(II) nitrate hexahydrate (Co(NO3)2·6H2O) is the hydrated cobalt salt. Its step-wise thermal degradation has been investigated. It affords anhydrous cobalt(II)nitrate as decomposition product.[1]

Application

Cobalt(II) nitrate hexahydrate may be used in the preparation of the following complexes (bz = benzimidazole, abz = albendazole, ntb = tris(2-benzimidazolylmethyl)amine, 2ab = 2-aminobenzimidazole):
  • dinitratobis-(triphenylarsine oxide)-cobalt(II) complex[2]
  • [Co(bz)2(NO3)2].0.5EtOH[3]
  • [Co(abz)2(NO3)2].2.5H2O[3]
  • [Co(ntb)NO3]NO3.H2O[3]

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Eye Dam. 1 - Muta. 2 - Ox. Sol. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 2 Inhalation

Target Organs

Lungs

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Some Phosphine Oxide and Arsine Oxide Complexes of Cobalt (II) Nitrate.
Cotton FA, et al.
Inorganic Chemistry, 2(6), 1162-1165 (1963)
Thermal decomposition of cobalt nitrato compounds: Preparation of anhydrous cobalt (II) nitrate and its characterisation by Infrared and Raman spectra.
Ehrhardt C, et al.
Thermochimica Acta, 432(1), 36-40 (2005)
Horacio López-Sandoval et al.
Journal of inorganic biochemistry, 102(5-6), 1267-1276 (2008-02-29)
In this work we present the synthesis, structural and spectroscopic characterisation of a series of cobalt(II) and zinc(II) coordination compounds with benzimidazole (bz) and its 2-benzimidazole derivatives: 2-aminobenzimidazole (2ab), albendazole (abz) and tris(2-benzimidazolylmethyl)amine (ntb). The compounds were evaluated for their
William Buchmann et al.
Journal of mass spectrometry : JMS, 42(4), 517-526 (2007-02-27)
The electronebulization of a cobalt(II)/cysteine(Cys) mixture in water/methanol (50/50) produced mainly cobalt-cationized species. Three main groups of the Co-cationized species can be distinguished in the ESI-MS spectrum: (1) the cobalt complexes including the cysteine amino acid only (they can be
N G Al-Tawil et al.
Acta dermato-venereologica, 64(3), 203-208 (1984-01-01)
Peripheral blood lymphocytes from 45 patients with a positive patch test to cobalt chloride (CoCl2) and 37 controls were stimulated with various concentrations of CoCl2 and/or cobalt sulphate (CoSO4) or cobalt nitrate (Co(NO3)2) or cobalt acetate [CH3COO)2Co) for various days

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service