Skip to Content
Merck
All Photos(1)

Key Documents

576913

Sigma-Aldrich

Water solution

suitable for HPLC, contains 0.1 % (v/v) formic acid

Synonym(s):

Aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

4 X 4 L
$723.00

$723.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
4 X 4 L
$723.00

About This Item

Linear Formula:
H2O
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
NACRES:
NA.21
grade:
HPLC grade
technique(s):
HPLC: suitable

$723.00


Check Cart for Availability

Request a Bulk Order

grade

HPLC grade

form

liquid

contains

0.1 % (v/v) formic acid

technique(s)

HPLC: suitable

SMILES string

O

InChI

1S/H2O/h1H2

InChI key

XLYOFNOQVPJJNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The product is water containing 0.1%v/v formic acid. This pre-blended mobile phase solvent is of high purity that reduces time-consuming preparation and prevents instrument downtime because of impure mobile phases. It can be employed for HPLC and UHPLC (Ultra High performance Liquid Chromatography) techniques.

Preparation Note

Product filtered through a 0.2 μm filter

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service