Skip to Content
Merck
All Photos(1)

Key Documents

320501

Sigma-Aldrich

Sulfuric acid

ACS reagent, 95.0-98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2SO4
CAS Number:
Molecular Weight:
98.08
Beilstein:
2037554
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

ACS reagent

vapor density

<0.3 (25 °C, vs air)

vapor pressure

1 mmHg ( 146 °C)

description

Free from suspended or insoluble matter
Nominally 95-98% H2SO4

Assay

95.0-98.0%

form

liquid

ign. residue

≤5 ppm

color

APHA: ≤10

pH

1.2 (5 g/L)

bp

~290 °C (lit.)

density

1.840 g/mL at 25 °C (lit.)

anion traces

MnO4- reducers: ≤2 ppm
chloride (Cl-): ≤0.2 ppm
nitrate (NO3-): ≤0.5 ppm

cation traces

As: ≤0.01 ppm
Fe: ≤0.2 ppm
Hg: ≤5 ppb
NH4+: ≤2 ppm
heavy metals (as Pb): ≤1 ppm

SMILES string

OS(O)(=O)=O

InChI

1S/H2O4S/c1-5(2,3)4/h(H2,1,2,3,4)

InChI key

QAOWNCQODCNURD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

João Almeida et al.
Nature, 502(7471), 359-363 (2013-10-08)
Nucleation of aerosol particles from trace atmospheric vapours is thought to provide up to half of global cloud condensation nuclei. Aerosols can cause a net cooling of climate by scattering sunlight and by leading to smaller but more numerous cloud
Francesco Riccobono et al.
Science (New York, N.Y.), 344(6185), 717-721 (2014-05-17)
Atmospheric new-particle formation affects climate and is one of the least understood atmospheric aerosol processes. The complexity and variability of the atmosphere has hindered elucidation of the fundamental mechanism of new-particle formation from gaseous precursors. We show, in experiments performed
Siegfried Schobesberger et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(43), 17223-17228 (2013-10-09)
Atmospheric aerosols formed by nucleation of vapors affect radiative forcing and therefore climate. However, the underlying mechanisms of nucleation remain unclear, particularly the involvement of organic compounds. Here, we present high-resolution mass spectra of ion clusters observed during new particle
Vicky De Preter et al.
Gut, 64(3), 447-458 (2014-05-09)
Bacteria play a role in the onset and perpetuation of intestinal inflammation in IBD. Compositional alterations may also change the metabolic capacities of the gut bacteria. To examine the metabolic activity of the microbiota of patients with Crohn's disease (CD)
I A Panagiotopoulos et al.
Bioresource technology, 130, 570-577 (2013-01-22)
A two-stage pretreatment approach, employing steam followed by organosolv treatment, was assessed for its ability to fractionate and recover most of the hemicellulose, lignin and cellulose components of poplar wood chips. A mild steaming stage was initially used to maximise

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service