Skip to Content
Merck
All Photos(1)

Key Documents

320145

Sigma-Aldrich

Ammonium hydroxide solution

ACS reagent, 28.0-30.0% NH3 basis

Synonym(s):

Ammonia aqueous, Ammonia water

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4OH
CAS Number:
Molecular Weight:
35.05
Beilstein:
3587154
MDL number:
UNSPSC Code:
12352106
eCl@ss:
38060103
PubChem Substance ID:

grade

ACS reagent

vapor density

1.2 (vs air)

vapor pressure

115 mmHg ( 20 °C)

form

liquid

expl. lim.

27 %

concentration

28.0-30.0% NH3

impurities

MnO4- reducers, passes test
≤0.002% CO2

ign. residue

≤0.002%

density

0.9 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.5 ppm
nitrate (NO3-): ≤2 ppm
phosphate (PO43-): ≤2 ppm
sulfate (SO42-): ≤2 ppm

cation traces

Fe: ≤0.2 ppm
heavy metals (as Pb): ≤0.5 ppm

SMILES string

[NH4+].[OH-]

InChI

1S/H3N.H2O/h1H3;1H2

InChI key

VHUUQVKOLVNVRT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Courtney R Sullivan et al.
Molecular neurobiology, 56(6), 4492-4517 (2018-10-20)
We utilized a cell-level approach to examine glycolytic pathways in the DLPFC of subjects with schizophrenia (n = 16) and control (n = 16) and found decreased mRNA expression of glycolytic enzymes in pyramidal neurons, but not astrocytes. To replicate these novel bioenergetic findings
Konstantin Chingin et al.
Analytical chemistry, 84(15), 6856-6862 (2012-07-12)
We introduce an online multiple-junction capillary isoelectric focusing fractionator (OMJ-CIEF) for separation of biological molecules in solution by pI. In OMJ-CIEF, the separation capillary is divided into seven equal sections joined with each other via tubular Nafion membrane insertions. Each
Enrique Cequier et al.
Analytica chimica acta, 845, 98-104 (2014-09-10)
Organophosphate triesters are common flame retardants used in a wide variety of consumer products from which they can migrate and pollute the indoor environment. Humans may thus be continuously exposed to several organophosphate triesters which might be a risk for
Aurélie Periat et al.
Journal of chromatography. A, 1356, 211-220 (2014-07-16)
In this study, the influence of electrospray ionization (ESI) source design on the overall sensitivity achieved in hydrophilic interaction chromatography (HILIC) and reversed phase liquid chromatography (RPLC), was investigated. State-of-the-art triple quadrupole mass analyzers from AB Sciex, Agilent Technologies and
Mohammed Anwar et al.
International journal of pharmaceutics, 468(1-2), 158-164 (2014-04-22)
Nano-sized curcumin-loaded super-paramagnetic iron oxide nanoparticles (CUR-OA-SPIONs) were synthesized chemically by co-precipitation method using oleic acid as a stabilizer and Myrj 52 as a surfactant. The synthesized nanoparticles were characterized for their shape, size, surface morphology, electrokinetic potential, magnetic properties

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service