Skip to Content
Merck
All Photos(3)

Key Documents

310514

Sigma-Aldrich

Sodium perchlorate monohydrate

ACS reagent

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$98.60
500 G
$278.00

$98.60


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 G
$98.60
500 G
$278.00

About This Item

Linear Formula:
NaClO4 · H2O
CAS Number:
Molecular Weight:
140.46
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
form:
powder, crystals or chunks

$98.60


Available to ship on22 April 2025Details


Request a Bulk Order

grade

ACS reagent

Quality Level

Assay

85.0-90.0% (monohydrate)

form

powder, crystals or chunks

reaction suitability

reagent type: oxidant

impurities

≤0.005% insolubles

pH

6.0-8.0 (25 °C, 5%)

anion traces

chloride (Cl-): ≤0.003%
sulfate (SO42-): ≤0.002%

cation traces

Ca: ≤0.02%
Fe: ≤5 ppm
K: ≤0.05%
heavy metals: ≤5 ppm (by ICP)

SMILES string

O.[Na+].[O-]Cl(=O)(=O)=O

InChI

1S/ClHO4.Na.H2O/c2-1(3,4)5;;/h(H,2,3,4,5);;1H2/q;+1;/p-1

InChI key

IXGNPUSUVRTQGW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium perchlorate monohydrate (NaClO4· H2O) is a hydrated sodium salt. Its crystals were reported to exhibit a monoclinic crystal system having space group C2/c. It is a strong oxidizer.

Application

Sodium perchlorate monohydrate can be:
  • Used as a supporting electrolyte in the preparation of polypyrrole (PPy) film.
  • Doped with host polymer NaCMC (sodium carboxymethyl cellulose) to prepare electrolyte film (NaCMC-NaClO4 H2O) which is used in the electrochemical cell.
  • Used as an oxidizing agent to remove color from the textiles waste water.

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 1

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Cynthia G Zoski et al.
Analytical chemistry, 79(13), 4957-4966 (2007-05-29)
We discuss SECM tip voltammetry, where a UME tip is held above a conductive substrate within about a tip radius and a tip voltammogram is recorded as its potential is slowly scanned while the substrate is held at a fixed
Hydrogen bond studies. CII. An X-ray determination of the crystal structure of sodium perchlorate monohydrate, NaClO4. H2O.
Berglund B, et al.
Acta Crystallographica Section B, Structural Science, 31(7), 1842-1846 (1975)
Deniz Ucar et al.
Environmental technology, 37(2), 183-191 (2015-06-24)
Nitrate and perchlorate were identified as significant water contaminants all over the world. This study aims at evaluating the performances of the heterotrophic-autotrophic sequential denitrification process for reductive nitrate and perchlorate removal from drinking water. The reduced nitrate concentration in
Christopher Polonyi et al.
Dalton transactions (Cambridge, England : 2003), 43(31), 11941-11949 (2014-06-27)
The ability of the anticancer active drug cisplatin to exert biological activity through interference with nucleic acid function is well documented. Since kinetics play a key role in determining product distributions in these systems, methods for accurate documentation of reactivity
Ana Vemić et al.
Journal of pharmaceutical and biomedical analysis, 102, 314-320 (2014-12-03)
The aim of this paper is to present a development of liquid chromatographic method when chaotropic salts are used as mobile phase additives following the QbD principles. The effect of critical process parameters (column chemistry, salt nature and concentration, acetonitrile

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service