Skip to Content
Merck
All Photos(2)

Key Documents

308277

Sigma-Aldrich

Diethylene glycol diethyl ether

suitable for HPLC, ≥99%

Synonym(s):

2-Ethoxyethyl ether, Bis(2-ethoxyethyl) ether, Diethyldiglycol

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$502.00

$502.00


Available to ship on15 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 L
$502.00

About This Item

Linear Formula:
(C2H5OCH2CH2)2O
CAS Number:
Molecular Weight:
162.23
Beilstein:
1699259
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
NACRES:
NA.21
grade:
HPLC grade
technique(s):
HPLC: suitable
application(s):
food and beverages
bp:
180-190 °C (lit.)
vapor pressure:
0.5 mmHg ( 20 °C)

$502.00


Available to ship on15 April 2025Details


Request a Bulk Order

grade

HPLC grade

vapor density

5.6 (vs air)

vapor pressure

0.5 mmHg ( 20 °C)

Assay

≥99%

form

liquid

purified by

glass distillation

expl. lim.

6.2 %

technique(s)

HPLC: suitable

impurities

<0.050% water

refractive index

n20/D 1.412 (lit.)

bp

180-190 °C (lit.)

mp

-68 °C

density

0.909 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 260 nm Amax: 1.00
λ: 300 nm Amax: 0.10
λ: 320 nm Amax: 0.04
λ: 350-400 nm Amax: 0.01

application(s)

food and beverages

SMILES string

CCOCCOCCOCC

InChI

1S/C8H18O3/c1-3-9-5-7-11-8-6-10-4-2/h3-8H2,1-2H3

InChI key

RRQYJINTUHWNHW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

152.6 °F - closed cup

Flash Point(C)

67 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R L Schuler et al.
Environmental health perspectives, 57, 141-146 (1984-08-01)
Fifteen glycol ethers were investigated for their potential to cause adverse reproductive toxic effects using an in vivo mouse screening bioassay. Pregnant mice were orally dosed once per day on days 7 through 14 of gestation at concentrations causing 0
Excess molar volumes and viscosities of binary mixtures of diethylene glycol diethyl ether with chloroalkanes at 298.15 K.
Pal A and Bhardwaj RK.
Journal of Molecular Liquids, 102(1), 197-211 (2003)
Density, viscosity, and surface tension at 293.15 K and liquid-liquid equilibria from 301.15 K to 363.15 K under atmospheric pressure for the binary mixture of diethylene glycol diethyl ether+ water.
Li XX, et al.
Journal of Chemical and Engineering Data, 49(4), 1043-1045 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service