Skip to Content
Merck
All Photos(1)

Key Documents

30411

Sigma-Aldrich

Sodium pyrophosphate tetrabasic decahydrate

puriss. p.a., ≥99%

Synonym(s):

Pyrophosphate inorganic, Sodium diphosphate tetrabasic, Tetrasodium pyrophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na4P2O7 · 10H2O
CAS Number:
Molecular Weight:
446.06
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
form:
solid

grade

puriss. p.a.

Assay

≥99%

form

solid

impurities

≤0.0005% heavy metals (as Pb)
≤0.001% total nitrogen (N)

anion traces

chloride (Cl-): ≤10 mg/kg
phosphate (PO43-): ≤1000 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

As: ≤1 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg

SMILES string

O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[Na+].[O-]P([O-])(=O)OP([O-])([O-])=O

InChI

1S/4Na.H4O7P2.10H2O/c;;;;1-8(2,3)7-9(4,5)6;;;;;;;;;;/h;;;;(H2,1,2,3)(H2,4,5,6);10*1H2/q4*+1;;;;;;;;;;;/p-4

InChI key

VZWGHDYJGOMEKT-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shafigh Shafie et al.
Fish & shellfish immunology, 82, 115-120 (2018-08-10)
Adjuvant effect of G2 (extraction of buffalo spleen) was assessed in intraperitoneally immunized rainbow trout (100-150 g) with killed- Y. ruckeri bacterin biotype I [0.1 mL (1 × 107 cells/fish) of vaccine diluted with the adjuvant in a ratio of 1:1 (V/V)] at 12.4 ± 1.3 °C for
Cecilia C Low Wang et al.
Journal of diabetes and its complications, 27(4), 307-315 (2013-04-06)
Metabolic syndrome affects a large proportion of the population and increases cardiovascular disease risk. Because metabolic syndrome often co-exists clinically with atherosclerosis, it is difficult to distinguish the respective contributions of the components to vascular abnormalities. Accordingly, we utilized a
Shaefali P Rodgers et al.
Endocrinology, 151(3), 1194-1203 (2010-01-14)
We determined whether transient exposure to estradiol during middle age in ovariectomized rats would exert lasting effects on cognition and the brain beyond the period of exposure. Two experiments were conducted. Rats 10-11 months of age were ovariectomized and received
J Bohacek et al.
Journal of neuroendocrinology, 20(8), 1023-1027 (2008-06-11)
The role of oestrogen replacement therapy in preventing or delaying age-associated cognitive decline is controversial. Therapy success may critically depend on the time of treatment initiation following cessation of ovarian function. The present study aimed to assess, in middle-aged rats
Matthew Prideaux et al.
PloS one, 7(5), e36786-e36786 (2012-05-16)
Osteocytes are terminally differentiated osteoblasts which reside in a mineralized extracellular matrix (ECM). The factors that regulate this differentiation process are unknown. We have investigated whether ECM mineralization could promote osteocyte formation. To do this we have utilised MLO-A5 pre-osteocyte-like

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service