Skip to Content
Merck
All Photos(1)

Key Documents

289825

Sigma-Aldrich

Aluminum acetate, basic

Synonym(s):

Aluminum diacetate hydroxide, Aluminum hydroxide acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al(OH)(C2H3O2)2
CAS Number:
Molecular Weight:
162.08
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.55

form

powder

SMILES string

CC(=O)O[Al](O)OC(C)=O

InChI

1S/2C2H4O2.Al.H2O/c2*1-2(3)4;;/h2*1H3,(H,3,4);;1H2/q;;+3;/p-3

InChI key

HQQUTGFAWJNQIP-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Physical form

Insoluble form

Other Notes

No stabilizers added.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Alumina Powders from Aluminum Alkoxide.
Ayral A, et al.
MRS Proceedings, 121 (1988)
Alumina powders via a controlled precipitation of aluminum acetate.
Ayral A and Droguet JC.
Journal of Materials Research, 4(04), 967-971 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service