Skip to Content
Merck
All Photos(1)

Key Documents

237159

Sigma-Aldrich

Lead subacetate

ACS reagent

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pb(C2H3O2)2 · 2Pb[OH]2
CAS Number:
Molecular Weight:
807.72
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

ACS reagent

reaction suitability

reagent type: catalyst
core: lead

impurities

≤0.02% insol. dil. HOAc
≤1.0% insol. H2O
≥33.0% basic lead

loss

≤1.5% loss on drying

anion traces

NO3- and NO2-: passes test
chloride (Cl-): ≤0.003%

cation traces

Cu: ≤0.002%
Fe: ≤0.002%
K: ≤0.02%
Na: ≤0.05%

SMILES string

O[PbH2]O.O[PbH2]O.CC(=O)O[PbH2]OC(C)=O

InChI

1S/2C2H4O2.4H2O.3Pb/c2*1-2(3)4;;;;;;;/h2*1H3,(H,3,4);4*1H2;;;/q;;;;;;3*+2/p-6

InChI key

VLOJXAQYHIVPFI-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Repr. 1A - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Tülin Mansur et al.
Mycoses, 54(4), e205-e207 (2009-11-03)
Fusarium species may cause localised skin infections in immunocompetent individuals. At least half of these infections are preceded by skin breakdown. The lesions are characterised by slow progression and good response to therapy. Here we present a 60-year-old non-diabetic man
F J Fritz et al.
International archives of allergy and applied immunology, 88(3), 360-362 (1989-01-01)
Normal adult rats were used to quantitate and characterize mast cells in the male genital tract. The tissues were either fixed in a fixative containing formalin (Schaffer solution) or with basic lead acetate (BLA) to identify 'connective-tissue mast cells' and
L A Poirier et al.
Cancer research, 44(4), 1520-1522 (1984-04-01)
The ability of the physiologically essential divalent metals calcium and magnesium to inhibit the tumorigenic activities of lead and nickel towards the lungs of strain A mice was investigated. The tumorigenic salts lead(II) subacetate and nickel(II) acetate were injected i.p.
D Befus et al.
International archives of allergy and applied immunology, 76(3), 232-236 (1985-01-01)
We have studied the distribution and abundance of histochemically distinct mast cell subpopulations in the normal small and large intestine of man. One subpopulation has histochemical properties similar to the functionally distinct intestinal mucosal mast cell (MMC) of the rat
K S Kasprzak et al.
Carcinogenesis, 6(2), 279-282 (1985-02-01)
Seven groups of 4-week-old male Sprague-Dawley rats, 30 animals/group, were fed for 18 months a commercial chow diet with or without 1% admixture of lead subacetate (Pb-Acet) and 0, 0.3, 1, 3 and 6% of calcium acetate (CaAcet). Feeding a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service