Skip to Content
Merck
All Photos(2)

Key Documents

228702

Sigma-Aldrich

Sodium metaborate hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$63.30

$63.30


Estimated to ship on18 May 2025


Request a Bulk Order

Select a Size

Change View
500 G
$63.30

About This Item

Linear Formula:
NaBO2 · xH2O
CAS Number:
Molecular Weight:
65.80 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
for analytical purposes
form:
powder or crystals (or granules)

$63.30


Estimated to ship on18 May 2025


Request a Bulk Order

grade

for analytical purposes

form

powder or crystals (or granules)

concentration

7.7-10.7% NaOH (titration)

SMILES string

[Na+].[H]O[H].[O-]B=O

InChI

1S/BO2.Na.H2O/c2-1-3;;/h;;1H2/q-1;+1;

InChI key

UYAQHLFAIDLZLE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Minimizing water utilization in hydrolysis of sodium borohydride: the role of sodium metaborate hydrates.
Marrero-Alfonso EY, et al.
International Journal of Hydrogen Energy, 32(18), 4723-4730 (2007)
Synthesis, crystal structure and dehydration kinetics of NaB(OH)4?2H2O.
Kanturk A, et al.
Korean Journal of Chemical Engineering, 25(6), 1331-1337 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service