Skip to Content
Merck
All Photos(1)

Key Documents

12047

Sigma-Aldrich

Calcium oxide

puriss., meets analytical specification of FCC, powder (fine)

Synonym(s):

Lime, Quicklime

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaO
CAS Number:
Molecular Weight:
56.08
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.21
form:
powder (fine)

grade

puriss.

form

powder (fine)

quality

meets analytical specification of FCC

reaction suitability

core: calcium

impurities

≤0.5% insoluble in HCl
≤2.5% Mg and alkali salts (SO4)

loss

≤3.5% loss on ignition

pH

12.6 (20 °C)

bp

2850 °C (lit.)

density

3.3 g/mL at 25 °C (lit.)

anion traces

fluoride (F-): ≤150 mg/kg

cation traces

As: ≤3 mg/kg
Pb: ≤2 mg/kg

storage temp.

−20°C

SMILES string

O=[Ca]

InChI

1S/Ca.O

InChI key

ODINCKMPIJJUCX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of biodiesel catalyzed by solid super base of calcium oxide and its refining process.
Huaping ZHU, et al.
Chinese Journal of Catalysis, 27(5), 391-396 (2006)
Calcium oxide as a solid base catalyst for transesterification of soybean oil and its application to biodiesel production.
Kouzu M, et al.
Fuel: The Science and Technology of Fuel and Energy, 87(12), 2798-2806 (2008)
Marco Faustini et al.
Nanoscale, 5(3), 984-990 (2012-12-19)
We report the simple preparation of ultra-thin self-assembled nanoperforated titanium calcium oxide films and their use as reactive nanomasks for selective dry etching of silicon. This novel reactive nanomask is composed of TiO(2) in which up to 50% of Ti
M Lopes et al.
Poultry science, 92(3), 638-644 (2013-02-26)
Testing different management practices can help to identify conditions that decrease or even eliminate pathogenic bacteria in poultry litter. A trial was conducted to evaluate the effects of daily manual stirring (rotation of the litter with a pitchfork) for the
E Belon et al.
The Science of the total environment, 439, 87-95 (2012-10-16)
The inputs of ten trace elements (As, Cd, Cu, Cr, Hg, Mo, Ni, Pb, Se, Zn) to French agricultural soils have been assessed. The six main sources considered were: pesticides, mineral fertilizers, animal manure, liming materials, sludge and composts and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service