Skip to Content
Merck
All Photos(1)

Documents

11028

Sigma-Aldrich

Aluminum oxide

puriss., ≥98% Al2O3 basis

Synonym(s):

Alumina

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al2O3
CAS Number:
Molecular Weight:
101.96
EC Number:
MDL number:
UNSPSC Code:
12352311
eCl@ss:
38120402
PubChem Substance ID:

grade

puriss.

Assay

≥98% Al2O3 basis

form

solid

impurities

≤0.005% heavy metals (as Pb)
≤0.2% alkalis (as Na2O)

loss

≤0.5% loss on ignition, 1000 °C, 2 h

mp

2040 °C (lit.)

anion traces

chloride (Cl-): ≤50 mg/kg
silicate (as SiO2): ≤1000 mg/kg
sulfate (SO42-): ≤500 mg/kg

cation traces

Fe: ≤300 mg/kg

SMILES string

O=[Al]O[Al]=O

InChI

1S/2Al.3O

InChI key

TWNQGVIAIRXVLR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Aluminum oxide (Alumina, Al2O3) is commonly used as adsorbent. Formation of protective α-Al2O3 scales on the alloys at high temperatures has been studied. Polycrystalline Al2O3 has been employed to investigate the self-diffusion coefficient of aluminum in the temperature range of 1670°-1905°C. Aluminium oxide can either be obtained via thermal conversion of aluminium hydroxide at temperatures >1200οC or via combustion of aluminium. Aluminium oxide nanoparticles can behave like safe adjuvants and carriers for eliciting antibodies from small molecules called haptens.

Application

Aluminum oxide (Alumina) may be employed for the selective adsorption of the catecholamines during their determination in diverse biological matrices. It may be used in the fabrication of polycrystalline lithium iodide ­- aluminum oxide solid electrolytes. The incorporation of aluminum oxide remarkably increased the conductivity of lithium iodide. Aluminum oxide (activity II-III, according to the Brockmann scale) has been used for the chromatographic purifications.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eastaugh N, et al
Pigment Compendium: A Dictionary and Optical Microscopy of Historical Pigments, By Nicholas Eastaugh, Valentine Walsh, Tracey Chaplin (2008)
A study of the factors affecting the aluminum oxide-trihydroxyindole procedure for the analysis of catecholamines.
A H ANTON et al.
The Journal of pharmacology and experimental therapeutics, 138, 360-375 (1962-12-01)
"Aluminum oxide nanoparticles as carriers and adjuvants for eliciting antibodies from non-immunogenic haptens"
Maquieira A, et al.
Analytical Chemistry, 84(21), 9340-9348 (2012)
Zhi-Yu Chen et al.
Polymers, 12(11) (2020-11-04)
Tendon healing after injury is relatively slow, mainly because of the weak activity and metabolic properties of tendon cells (tenocytes). Bletilla striata polysaccharide (BSP) has been reported to enhance cell proliferation. Here, we aimed to increase tendon cell proliferation by
Valentina La Rosa et al.
Antimicrobial agents and chemotherapy, 56(1), 324-331 (2011-10-26)
The 1,5-diarylpyrrole derivative BM212 was previously shown to be active against multidrug-resistant clinical isolates and Mycobacterium tuberculosis residing within macrophages as well as against Mycobacterium avium and other atypical mycobacteria. To determine its mechanism of action, we identified the cellular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service