Skip to Content
Merck
All Photos(1)

Key Documents

10317

Sigma-Aldrich

Bismuth(III) carbonate basic

puriss., meets analytical specification of Ph. Eur., 80-82.5% Bi basis (calc. on dried substance)

Synonym(s):

Bismuth subcarbonate

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$84.60

$84.60


Available to ship on11 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 G
$84.60

About This Item

Linear Formula:
(BiO)2CO3
CAS Number:
Molecular Weight:
509.97
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21
form:
powder or solid

$84.60


Available to ship on11 April 2025Details


Request a Bulk Order

grade

puriss.

Assay

80-82.5% Bi basis (calc. on dried substance)

form

powder or solid

quality

meets analytical specification of Ph. Eur.

impurities

residual solvents, complies
≤1% alkalis/earth alkalis (as SO4)

loss

≤1% loss on drying, 105 °C

density

6.86 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤500 mg/kg
nitrate (NO3-): ≤4000 mg/kg

cation traces

Ag: ≤20 mg/kg
As: ≤5 mg/kg
Cu: ≤50 mg/kg
Pb: ≤20 mg/kg

suitability

complies for appearance of solution

SMILES string

O=[Bi]OC(=O)O[Bi]=O

InChI

1S/CH2O3.2Bi.2O/c2-1(3)4;;;;/h(H2,2,3,4);;;;/q;2*+1;;/p-2

InChI key

MGLUJXPJRXTKJM-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Experimental studies on altering the properties of the prosthetic plastic, Kallocryl B, by adding an x-ray contrast medium].
R Deutschmann et al.
Stomatologie der DDR, 30(7), 475-481 (1980-07-01)
Fan Dong et al.
Dalton transactions (Cambridge, England : 2003), 41(27), 8270-8284 (2012-05-25)
The synthesis and self-assembly of hierarchical architectures from nanoscale building blocks with unique morphology, orientation and dimension have opened up new opportunities to enhance their functional performances and remain a great challenge. This work represents tunable synthesis of various types
Comparison of plasma bismuth levels after oral dosing with basic bismuth carbonate or tripotassium dicitrato bismuthate.
Madaus S, et al.
Alimentary Pharmacology & Therapeutics, 6(2), 241-249 (1992)
Thomas Serena et al.
Advances in skin & wound care, 20(9 Pt 1), 485-492 (2007-09-01)
The human forearm biopsy model can evaluate the effect of novel agents on acute wounds. Bismuth subgallate/borneol (Suile) is a new product cleared by the Food and Drug Administration for treating partial-thickness wounds. Anecdotal reports suggest that Suile may be
Rong Chen et al.
Chemical communications (Cambridge, England), (21)(21), 2265-2267 (2006-05-24)
Bismuth subcarbonate ((BiO)2CO3) nanotubes with uniform diameters of about 3-5 nm were fabricated from bismuth citrate; these nanotubes exhibit antibacterial properties against Helicobacter pylori (50% inhibition at 10 microg mL(-1)), a bacterium causing peptic ulcers and gastritis.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service