Skip to Content
Merck
All Photos(1)

Key Documents

Y0001969

Etanercept

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$265.00

$265.00


Available to ship on14 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 MG
$265.00

About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24

$265.00


Available to ship on14 April 2025Details


Request a Bulk Order

grade

pharmaceutical primary standard

API family

etanercept

manufacturer/tradename

EDQM

application(s)

pharmaceutical

format

neat

Looking for similar products? Visit Product Comparison Guide

General description

Etanercept is one of the biologic disease-modifying anti-rheumatic drugs (DMARDs) used to treat rheumatoid arthritis (RA). It acts as a tumor necrosis factor (TNF) antagonist by binding with TNF cytokine or protein, thereby blocking its interaction with cell surface receptors. Thus, etanercept effectively relieves joint inflammation from RA and lowers the disease activity.[1][2][3][4][5][6]
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

This product is intended for laboratory tests as prescribed in the EP only. Established for use with the monograph(s): 2895.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of a sensitive enzyme-linked immunosorbent assay for the measurement of biologically active etanercept in patients with ankylosing spondylitis
Wang L, et al.
Journal of Chromatography. B, Biomedical Applications, 1008, 219-224 (2016)
Etanercept
European Pharmacopoeia Commission and Directorate for the Quality of Medicines & Healthcare
European pharmacopoeia, 4979-4983 (2021)
Etanercept concentration in patients with rheumatoid arthritis and its potential influence on treatment decisions: a pilot study
Daien CI, et al.
The Journal of Rheumatology, 39(8), 1533-1538 (2012)
Etanercept: An overview
Goffe B and Cather JC
Journal of the American Academy of Dermatology, 49(2), 105-111 (2003)
Characterization and application of a unique panel of monoclonal antibodies generated against etanercept
Detrez I, et al.
Journal of Immunology, 196(6), 2879-2884 (2016)

Questions

1–2 of 2 Questions  
  1. Bonjour, j'ai reconstitué un aliquot d'étanercept dans l'eau. combien de temps puis je le garder à 4 degres ou est il preferable de le congeler?

    1 answer
    1. Recommendations against the long-term storage and use of this standard are listed in the associated Information Leaflet published by the European Pharmacopoeia. From the leaflet:

      "The container should not be opened until required for use. Allow the closed container to equilibrate at ambient temperature before opening to avoid uptake of moisture. This substance has been freeze-dried and is known to be hygroscopic. Tap the container gently to collect the material at the bottom. Reconstitute the contents by injecting a suitable volume of the solvent. Mix gently to ensure complete dissolution. When necessary transfer the solution to an appropriate volumetric flask and dilute to the required concentration. Ph. Eur. RS are for immediate use. Once the container has been opened, its entire content must be used immediately. Any further storage and re-use are not warranted."

      Helpful?

  2. What is the recommended method for reconstituting Etanercept (Y0001969), and how should the stock solutions be stored?

    1 answer
    1. Etanercept (Y0001969) is soluble in water. Once reconstituted, stock solutions should be aliquoted into tightly sealed vials and used within one month to ensure stability. It is important to avoid repeated freeze-thaw cycles to maintain the integrity of the solution.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service