This is a product of the European Pharmacopoeia. The solubility of this product is not determined. However, the research grade product option, Q3001, is tested for solubility at 25 mg/mL in Ethanol. In addition, various sources state that quercitrin be soluble in DMSO and DMF at 30 mg/ml.
Y0001931
Quercitrin
European Pharmacopoeia (EP) Reference Standard
Synonym(s):
3,3′,4′,5,7-Pentahydroxyflavone 3-rhamnoside, 3-O-Rhamnosylquercetin, Quercetin 3-rhamnoside, Quercimelin, Quercitroside
Select a Size
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
quercitrin
manufacturer/tradename
EDQM
format
neat
storage temp.
2-8°C
SMILES string
C[C@@H]1O[C@@H](OC2=C(Oc3cc(O)cc(O)c3C2=O)c4ccc(O)c(O)c4)[C@H](O)[C@H](O)[C@H]1O
InChI
1S/C21H20O11/c1-7-15(26)17(28)18(29)21(30-7)32-20-16(27)14-12(25)5-9(22)6-13(14)31-19(20)8-2-3-10(23)11(24)4-8/h2-7,15,17-18,21-26,28-29H,1H3/t7-,15-,17+,18+,21-/m0/s1
InChI key
OXGUCUVFOIWWQJ-HQBVPOQASA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Packaging
Other Notes
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
cas number:522-12-3 제품을 어떤 용매에 녹여야할까요? solvent 가 작성 되어있지 않아 질문드립니다.
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service