Skip to Content
Merck
All Photos(1)

Key Documents

Y0001740

Buserelin for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Buserelin acetate salt, des-Gly10-[D-Ser[t-Bu]6]-LH-RH ethylamide acetate salt, pGlu-His-Trp-Ser-Tyr-D-Ser(t-Bu)-Leu-Arg-Pro-NHEt acetate salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$218.00

$218.00


Estimated to ship on22 March 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$218.00

About This Item

Empirical Formula (Hill Notation):
C60H86N16O13 · C2H4O2
CAS Number:
Molecular Weight:
1299.48
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$218.00


Estimated to ship on22 March 2025


Request a Bulk Order

grade

pharmaceutical primary standard

API family

buserelin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

[nH]1cncc1C[C@H](NC(=O)[C@H]6NC(=O)CC6)C(=O)N[C@@H](Cc4c5c([nH]c4)cccc5)C(=O)N[C@@H](CO)C(=O)N[C@@H](Cc3ccc(cc3)O)C(=O)N[C@H](COC(C)(C)C)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCCN=C(N)N)C(=O)N2[C@@H](CCC2)C(=O)NCC.OC(=O)C

InChI

1S/C60H86N16O13.C2H4O2/c1-7-64-57(87)48-15-11-23-76(48)58(88)41(14-10-22-65-59(61)62)69-51(81)42(24-33(2)3)70-56(86)47(31-89-60(4,5)6)75-52(82)43(25-34-16-18-37(78)19-17-34)71-55(85)46(30-77)74-53(83)44(26-35-28-66-39-13-9-8-12-38(35)39)72-54(84)45(27-36-29-63-32-67-36)73-50(80)40-20-21-49(79)68-40;1-2(3)4/h8-9,12-13,16-19,28-29,32-33,40-48,66,77-78H,7,10-11,14-15,20-27,30-31H2,1-6H3,(H,63,67)(H,64,87)(H,68,79)(H,69,81)(H,70,86)(H,71,85)(H,72,84)(H,73,80)(H,74,83)(H,75,82)(H4,61,62,65);1H3,(H,3,4)/t40-,41-,42-,43-,44-,45-,46-,47+,48-;/m0./s1

InChI key

PYMDEDHDQYLBRT-DRIHCAFSSA-N

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Glp-His-Trp-Ser-Tyr-Ser-tBu-Leu-Arg-Pro-NHEt

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Buserelin for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service