Skip to Content
Merck
All Photos(1)

Key Documents

Y0001653

Chlormadinone acetate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

17α-Acetoxy-6-chloro-4,6-pregnadiene-3,20-dione, 6-Chloro-17α-hydroxy-4,6-pregnadiene-3,20-dione 17-acetate, 6-Chloro-6-dehydro-17α-acetoxyprogesterone, Gestafortin, Matrol, Menstridyl

Sign Into View Organizational & Contract Pricing

Select a Size

130 MG
$254.00

$254.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
130 MG
$254.00

About This Item

Empirical Formula (Hill Notation):
C23H29ClO4
CAS Number:
Molecular Weight:
404.93
Beilstein:
2633614
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$254.00


Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

fraxinus chinensis

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[H][C@]12CC[C@@]3(C)[C@@]([H])(CC[C@]3(OC(C)=O)C(C)=O)[C@]1([H])C=C(Cl)C4=CC(=O)CC[C@]24C

InChI

1S/C23H29ClO4/c1-13(25)23(28-14(2)26)10-7-18-16-12-20(24)19-11-15(27)5-8-21(19,3)17(16)6-9-22(18,23)4/h11-12,16-18H,5-10H2,1-4H3/t16-,17+,18+,21-,22+,23+/m1/s1

InChI key

QMBJSIBWORFWQT-DFXBJWIESA-N

Gene Information

human ... PGR(5241)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Chlormadinone acetate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Review of progestogenic and related pharmacological effects of chlormadinone acetate.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Bouchard
The European journal of contraception & reproductive health care : the official journal of the European Society of Contraception, 10 Suppl 1, 7-11 (2005-12-17)
Chlormadinone acetate (CMA) is a derivative of naturally secreted progesterone that shows high affinity and activity at the progesterone receptor. It has an anti-estrogenic effect and, in contrast to natural progesterone, shows moderate anti-androgenic properties. CMA acts by blocking androgen
Seiji Furuya et al.
Hinyokika kiyo. Acta urologica Japonica, 51(3), 159-164 (2005-04-28)
Between August 1985 and March 2004, we performed transurethral resection of the prostate (TURP) in 18 patients with benign prostatic hyperplasia (BPH) whose prostatic volume was larger than 100 ml. We divided the patients into two groups. Group A consisted
Aurora Guerra-Tapia et al.
American journal of clinical dermatology, 12 Suppl 1, 3-11 (2011-09-16)
Acne vulgaris, hirsutism, seborrhea and female pattern hair loss (FPHL) are common disorders of the pilosebaceous unit (PSU). In some women with hyperandrogenemia, an excess of androgens at the PSU can lead to the development of these dermatological manifestations. These
Hideyuki Iwahata et al.
Journal of assisted reproduction and genetics, 32(4), 521-526 (2015-01-27)
Closed vitrification poses a risk of adversely affecting embryo development, while it may minimize the risk of contamination. We assessed the effects of closed-system human embryo vitrification on fetal development after implantation, neonatal outcome, and clinical safety. This was a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service