Skip to Content
Merck
All Photos(1)

Key Documents

Y0001611

Esculetin

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

6,7-Dihydroxycoumarin, Cichorigenin, Esculetin

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$221.00

$221.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$221.00

About This Item

Empirical Formula (Hill Notation):
C9H6O4
CAS Number:
Molecular Weight:
178.14
Beilstein:
152788
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$221.00


Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

esculetin

manufacturer/tradename

EDQM

mp

271-273 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Oc1cc2OC(=O)C=Cc2cc1O

InChI

1S/C9H6O4/c10-6-3-5-1-2-9(12)13-8(5)4-7(6)11/h1-4,10-11H

InChI key

ILEDWLMCKZNDJK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Esculetin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Rashi Arora et al.
Molecular cancer, 15(1), 64-64 (2016-10-21)
A handful of studies have exploited antitumor potential of esculetin, a dihydroxy coumarine derivative; the targets to which it binds and the possible downstream mechanism for its cytotoxicity in cancer cells remain to be elucidated. Using pancreatic cancer cell lines
Edson Luis Maistro et al.
Journal of toxicology and environmental health. Part A, 78(2), 109-118 (2014-11-27)
Coumarins are naturally occurring compounds, widely distributed throughout the plant kingdom (Plantae), and possess important pharmacological properties, including inhibition of oxidative stress. In this context, newly synthesized coumarin compounds are being produced due to their potent antioxidant activities. Therefore, the
Hisayo Yamaoka et al.
Plastic and reconstructive surgery, 134(1), 50e-58e (2014-07-17)
Injectable collagen is often used for treatment of wrinkles or scars in cosmetic surgery. However, it is degraded within a short period after subcutaneous injection. The authors aimed to achieve a long-lasting effect of the filler with a new collagenase
Yoon-Young Sung et al.
Environmental toxicology and pharmacology, 38(1), 41-50 (2014-06-01)
Viola mandshurica W. Becker has been used as an expectorant, diuretic, and anti-inflammatory agent. We evaluated the effects of V. mandshurica ethanol extract (VME) on high-fat-diet (HFD)-induced obesity in mice. HPLC analysis showed that the VME contained 11.95 ± 0.37
Oussama Ahrazem et al.
Plant science : an international journal of experimental plant biology, 234, 60-73 (2015-03-26)
Glycosyltransferases play diverse roles in cellular metabolism by modifying the activities of regulatory metabolites. Three stress-regulated UDP-glucosyltransferase-encoding genes have been isolated from the stigmas of saffron, UGT85U1, UGT85U2 and UGT85V1, which belong to the UGT85 family that includes members associated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service