Skip to Content
Merck
All Photos(1)

Key Documents

Y0001555

Ciclesonide impurity B

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Desisobutyryl-ciclesonide, (2′R)-2′-Cyclohexyl-11b,21-dihydroxy-16bH-[1,3]dioxolo[4′,5′:16,17]pregna-1,4-diene-3,20-dione

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$319.00

$319.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$319.00

About This Item

Empirical Formula (Hill Notation):
C28H38O6
CAS Number:
Molecular Weight:
470.60
UNSPSC Code:
41116107
NACRES:
NA.24

$319.00


Check Cart for Availability

Request a Bulk Order

biological source

synthetic

grade

pharmaceutical primary standard

Agency

EP

API family

ciclesonide

form

powder

packaging

pkg of 10 mg

manufacturer/tradename

EDQM

storage condition

protect from light

solubility

water: <0.1 g/L

application(s)

pharmaceutical (small molecule)

format

neat

shipped in

ambient

storage temp.

2-8°C

SMILES string

O1[C@@]2([C@H](O[C@H]1C6CCCCC6)C[C@@H]3[C@@]2(C[C@@H]([C@H]4[C@H]3CCC5=CC(=O)C=C[C@@]54C)O)C)C(=O)CO

InChI

1S/C28H38O6/c1-26-11-10-18(30)12-17(26)8-9-19-20-13-23-28(22(32)15-29,27(20,2)14-21(31)24(19)26)34-25(33-23)16-6-4-3-5-7-16/h10-12,16,19-21,23-25,29,31H,3-9,13-15H2,1-2H3/t19-,20-,21-,23+,24+,25+,26-,27-,28+/m0/s1

InChI key

OXPLANUPKBHPMS-ZXBNPROVSA-N

General description

Ciclesonide impurity B is an impurity of ciclesonide, a new-generation, non-halogenated glucocorticoid.[1][2][3]
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ciclesonide impurity B is used as an EP reference standard to quantify the analyte in pharmaceutical formulations by liquid chromatography (LC) technique.[4]

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New Drugs for Asthma, Allergy and COPD, 42, 42-54 (2001)
Ciclesonide
European Pharmacopoeia Commission and European Directorate for the Quality of Medicines & Healthcare
European pharmacopoeia, 43, 2206-2207 (2017)
Rethinking Cleaning Validation for API Manufacturing
Zhang C, et al.
Pharmaceutical Technology, 42, 42-54 (2018)
Pharmacokinetics of [14 C] ciclesonide after oral and intravenous administration to healthy subjects
Nave R, et al.
Clinical Pharmacokinetics, 43, 479-486 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service