Skip to Content
Merck
All Photos(1)

Key Documents

Y0001315

Josamycin for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Josamycin

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$204.00

$204.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$204.00

About This Item

Empirical Formula (Hill Notation):
C42H69NO15
CAS Number:
Molecular Weight:
827.99
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$204.00


Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

josamycin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CO[C@H]1[C@@H](CC(=O)O[C@H](C)C\C=C\C=C\[C@H](O)[C@H](C)C[C@H](CC=O)[C@@H]1O[C@@H]2O[C@H](C)[C@@H](O[C@H]3C[C@@](C)(O)[C@@H](OC(=O)CC(C)C)[C@H](C)O3)[C@@H]([C@H]2O)N(C)C)OC(C)=O

InChI

1S/C42H69NO15/c1-23(2)19-32(47)56-40-27(6)53-34(22-42(40,8)50)57-37-26(5)54-41(36(49)35(37)43(9)10)58-38-29(17-18-44)20-24(3)30(46)16-14-12-13-15-25(4)52-33(48)21-31(39(38)51-11)55-28(7)45/h12-14,16,18,23-27,29-31,34-41,46,49-50H,15,17,19-22H2,1-11H3/b13-12+,16-14+/t24-,25-,26-,27+,29+,30+,31-,34+,35-,36-,37-,38+,39+,40+,41+,42-/m1/s1

InChI key

XJSFLOJWULLJQS-NGVXBBESSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Josamycin for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Treatment of genital chlamydia infections with wilprafen (josamycin)].
L Damianov
Akusherstvo i ginekologiia, 42 Suppl 2, 35-37 (2003-11-19)
U Colombo et al.
Minerva ginecologica, 50(11), 491-497 (1999-01-12)
The macrolides are among the most effective antibiotics against infections due to Chlamydia and Mycoplasma. The drug in such cases must have marked antibacterial activity, good oral bioavailability, and high intracellular diffusion--indispensable for instance with Chlamydia infection. Rokitamycin, a macrolide
[A tick bite and lymphadenosis benigna cutis in childhood. Their treatment with josamycin].
I Querol Nasarre et al.
Anales espanoles de pediatria, 39(3), 251-254 (1993-09-01)
Shotaro Nonaka et al.
Journal of medical microbiology, 63(Pt 2), 242-247 (2013-11-08)
We investigated antimicrobial susceptibility and the molecular mechanism involved in conferring high-level macrolide resistance in 47 clinical isolates of Moraxella nonliquefaciens from Japan. Antimicrobial susceptibility was determined using Etest and agar dilution methods. Thirty-two erythromycin-non-susceptible strains were evaluated for the
Zhanhui Wang et al.
Food chemistry, 171, 98-107 (2014-10-14)
Erythromycin is an antibiotic used extensively in veterinary practice worldwide for treatment, prevention and growth promotion. In this work, monoclonal antibodies (Mabs) against erythromycin were produced and used to develop a competitive indirect enzyme-linked immunosorbent assay (ciELISA) for the determination

Questions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service