Skip to Content
Merck
All Photos(1)

Key Documents

Y0001183

Trolamine impurity B

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Diethanolamine, 2,2′-Iminodiethanol, Bis(2-hydroxyethyl)amine

Sign Into View Organizational & Contract Pricing

Select a Size

1.5 ML
$319.00

$319.00


Available to ship on14 April 2025Details


Request a Bulk Order

Select a Size

Change View
1.5 ML
$319.00

About This Item

Linear Formula:
HN(CH2CH2OH)2
CAS Number:
Molecular Weight:
105.14
Beilstein:
605315
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$319.00


Available to ship on14 April 2025Details


Request a Bulk Order

grade

pharmaceutical primary standard

vapor density

3.6 (vs air)

vapor pressure

<0.98 atm ( 100 °C)

API family

trolamine

autoignition temp.

689 °F

expl. lim.

10.6 %

manufacturer/tradename

EDQM

refractive index

n20/D 1.477 (lit.)

bp

217 °C/150 mmHg (lit.)

mp

28 °C (lit.)

density

1.097 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OCCNCCO

InChI

1S/C4H11NO2/c6-3-1-5-2-4-7/h5-7H,1-4H2

InChI key

ZBCBWPMODOFKDW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Trolamine impurity B EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Oral

Target Organs

Kidney,Liver,Blood

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

280.4 °F - closed cup

Flash Point(C)

138 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Honghui Yu et al.
Journal of immunology (Baltimore, Md. : 1950), 193(6), 2902-2910 (2014-08-15)
Anti-D can prevent immunization to the RhD Ag on RBCs, a phenomenon commonly termed Ab-mediated immune suppression (AMIS). The most accepted theory to explain this effect has been the rapid clearance of RBCs. In mouse models using SRBC, these xenogeneic
Hae-Rim Kim et al.
PloS one, 10(4), e0124909-e0124909 (2015-04-22)
Vascular endothelial growth factor (VEGF) has angiogenic, inflammatory, and bone-destructive roles in rheumatoid arthritis (RA). We aimed to determine the unique role of VEGF in osteoclastogenesis in RA. VEGF-induced receptor activator of nuclear factor ҡB ligand (RANKL) expression was determined
Hon-Wing Leung et al.
Regulatory toxicology and pharmacology : RTP, 43(3), 260-271 (2005-09-29)
Diethanolamine (DEA) is a chemical used widely in a number of industries and is present in many consumer products. Studies by the National Toxicology Program (NTP) have indicated that lifetime dermal exposure to DEA increased the incidence and multiplicity of
Choline deficiency associated with diethanolamine carcinogenicity.
Paul M Newberne
Toxicological sciences : an official journal of the Society of Toxicology, 67(1), 1-3 (2002-04-19)
M-L Metzger et al.
Clinical and experimental immunology, 179(2), 256-264 (2014-09-25)
Common variable immunodeficiency (CVID) encompasses a heterogeneous group of antibody deficiencies characterized by susceptibility to recurrent infections and sequelae, including bronchiectasis. We investigated the relevance of the lectin complement pathway in CVID patients by analysing ficolin-2 and ficolin-3 serum levels

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service