Skip to Content
Merck
All Photos(1)

Key Documents

Y0001088

Cyclopentolate for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Cyclopentolate hydrochloride, 2-Dimethylaminoethyl 1-hydroxy-α-phenylcyclopentaneacetate

Sign Into View Organizational & Contract Pricing

Select a Size

20 MG
$254.00

$254.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
20 MG
$254.00

About This Item

Empirical Formula (Hill Notation):
C17H25NO3 · HCl
CAS Number:
Molecular Weight:
327.85
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$254.00


Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

cyclopentolate

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl.CN(C)CCOC(=O)C(c1ccccc1)C2(O)CCCC2

InChI

1S/C17H25NO3.ClH/c1-18(2)12-13-21-16(19)15(14-8-4-3-5-9-14)17(20)10-6-7-11-17;/h3-5,8-9,15,20H,6-7,10-13H2,1-2H3;1H

InChI key

RHKZVMUBMXGOLL-UHFFFAOYSA-N

Gene Information

human ... CHRM3(1131)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Cyclopentolate for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yosuke Ito et al.
The Journal of veterinary medical science, 77(2), 155-160 (2014-11-07)
The purpose of this study was to investigate the effects of sevoflurane concentration on canine visual evoked potentials with pattern stimulation (P-VEPs). Six clinically normal laboratory-beagle dogs were used. The minimum alveolar concentration (MAC) of sevoflurane was detected from all
Zhen-Ying Cheng et al.
Graefe's archive for clinical and experimental ophthalmology = Albrecht von Graefes Archiv fur klinische und experimentelle Ophthalmologie, 252(12), 1939-1946 (2014-08-15)
To investigate the effects of the relatively selective GABAAOr receptor antagonist (1,2,5,6-tetrahydropyridin-4-yl) methylphosphinic acid (TPMPA) on form-deprivation myopia (FDM) in guinea pigs. A diffuser was applied monocularly to 30 guinea pigs from day 10 to 21. The animals were randomized
Yakov Goldich et al.
American journal of ophthalmology, 159(1), 155-159 (2014-12-03)
To compare objective and subjective outcomes after Descemet membrane endothelial keratoplasty (DMEK) and Descemet stripping automated endothelial keratoplasty (DSAEK) in the fellow eye of the same patients. Single-center, retrospective case series. Seventeen patients with bilateral Fuchs endothelial dystrophy who underwent
Jacky W Y Lee et al.
Medicine, 94(8), e567-e567 (2015-02-26)
To investigate the association between macular thickness and peripapillary retinal nerve fiber layer (RNFL) thickness in Chinese children. This cross-sectional study recruited consecutive cases of healthy pediatric subjects aged 4 to 18 from Caritas Medical Centre in Hong Kong Special
Marije J Hoornweg et al.
European journal of ophthalmology, 24(6), 940-947 (2014-04-15)
To compare intralesional corticosteroid (IC) injections with oral propranolol in children with periorbital infantile hemangioma (IH). Children were diagnosed with IH and treated with IC (n = 29) or propranolol (n = 14). In cases in which no further improvement

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service