Skip to Content
Merck
All Photos(1)

Key Documents

Y0000912

Paraffin (Yellow soft)

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Paraffin wax

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$262.00

$262.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$262.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$262.00


Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

paraffin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

InChI key

JWHAUXFOSRPERK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Paraffin (Yellow soft) EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Theresa M Mullan et al.
Forensic science, medicine, and pathology, 7(2), 198-208 (2010-09-23)
Among the various modes of death due to asphyxiation, drowning is unique. Unlike other types of suffocation associated with occluding airway obstructions (i.e. choking and smothering); with drowning, there is typically filling and involvement of the majority of the tracheobronchial
Antonio F Di Narzo et al.
Journal of the National Cancer Institute, 106(10), doi:10-doi:10 (2014-09-24)
Prognosis prediction for resected primary colon cancer is based on the T-stage Node Metastasis (TNM) staging system. We investigated if four well-documented gene expression risk scores can improve patient stratification. Microarray-based versions of risk-scores were applied to a large independent

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service