Skip to Content
Merck
All Photos(1)

Documents

Y0000838

Doxepin hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

11-(3-Dimethylaminopropylidene)-6,11-dihydrodibenz[b,e]oxepine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21NO · HCl
CAS Number:
Molecular Weight:
315.84
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

doxepin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl[H].CN(C)CC\C=C1/c2ccccc2COc3ccccc13

InChI

1S/C19H21NO.ClH/c1-20(2)13-7-11-17-16-9-4-3-8-15(16)14-21-19-12-6-5-10-18(17)19;/h3-6,8-12H,7,13-14H2,1-2H3;1H/b17-11+;

InChI key

MHNSPTUQQIYJOT-SJDTYFKWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Doxepin hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haramandeep Singh et al.
Expert opinion on investigational drugs, 16(8), 1295-1305 (2007-08-10)
Low-dose doxepin hydrochloride (1, 3 and 6 mg) is a tricyclic antidepressant currently being investigated for the treatment of primary insomnia in adult and geriatric patients. Although it has been used at much higher doses to treat depression effectively for
Feng Tao et al.
Journal of biochemical and molecular toxicology, 34(2), e22424-e22424 (2019-11-20)
Recent studies have shown that tricyclic antidepressants (TCAs) may have anti-inflammatory and anticonvulsant effects in addition to its antidepressant effects. So far, the nonantidepressant effects of TCAs and their molecular pharmacological mechanisms remain completely unclear. Chronic inflammation in the brain
Grzegorz Jagla et al.
Pharmacological reports : PR, 66(3), 459-465 (2014-06-07)
The therapy of neuropathic pain may include the use of co-analgesics, such as antidepressants, however, their desired analgesic effect is associated with significant side effects. An alternative approach to this is their local administration which has been proposed, but there
Alvaro Gimeno et al.
International journal of pharmaceutics, 477(1-2), 650-654 (2014-12-03)
According to previous studies reporting the anesthetic/analgesic action of oral topical doxepin administration, this study evaluated a model of buccal permeation to determine the depth of delivery of doxepin into excised porcine buccal mucosa following topical application of a saturated
Shigeru Hishinuma et al.
Biochemical pharmacology, 91(2), 231-241 (2014-07-30)
Differential binding sites for first- and second-generation antihistamines were indicated on the basis of the crystal structure of human histamine H1 receptors. In this study, we evaluated differences between the thermodynamic driving forces of first- and second-generation antihistamines for human

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service