Skip to Content
Merck
All Photos(1)

Key Documents

Y0000528

Sulbactam

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(2S,5R)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid 4,4-dioxide, Betamaze, CP 45899, CP-45,899, Penicillanic acid 1,1-dioxide, Penicillanic acid S,S-dioxide, Penicillanic acid dioxide, Penicillanic acid sulfone

Sign Into View Organizational & Contract Pricing

Select a Size

150 MG
$218.00

$218.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
150 MG
$218.00

About This Item

Empirical Formula (Hill Notation):
C8H11NO5S
CAS Number:
Molecular Weight:
233.24
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$218.00


Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

sulbactam

manufacturer/tradename

EDQM

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

CC1(C)[C@@H](N2[C@@H](CC2=O)S1(=O)=O)C(O)=O

InChI

1S/C8H11NO5S/c1-8(2)6(7(11)12)9-4(10)3-5(9)15(8,13)14/h5-6H,3H2,1-2H3,(H,11,12)/t5-,6+/m1/s1

InChI key

FKENQMMABCRJMK-RITPCOANSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sulbactam EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Serafim Tsitsilonis et al.
Langenbeck's archives of surgery, 398(1), 153-159 (2012-07-27)
Necrotizing fasciitis (NF) is a rare, but potentially fatal pathology. The aim of the present study was to identify the population characteristics of the NF patients, the responsible bacteria, and the differences between survivors and nonsurvivors. In this retrospective case-control
Elizabeth A Rodkey et al.
Journal of the American Chemical Society, 134(40), 16798-16804 (2012-09-15)
The rise of inhibitor-resistant and other β-lactamase variants is generating an interest in developing new β-lactamase inhibitors to complement currently available antibiotics. To gain insight into the chemistry of inhibitor recognition, we determined the crystal structure of the inhibitor preacylation
Bahadir M Gulluoglu et al.
Annals of surgery, 257(1), 37-43 (2012-09-25)
To assess the impact of prophylactic antibiotics on the prevention of surgical site infection (SSI) and the cost-effectiveness of this prophylaxis for breast cancer surgery in overweight or obese women. SSI is higher than expected after breast surgery. Obesity was
G S Singh
Mini reviews in medicinal chemistry, 4(1), 93-109 (2004-02-03)
Beta-lactam ring-containing compounds such as penicillins, ampicillin, amoxicillin, cephalosporins and carbapenems are among the most famous antibiotics. This article reviews the recent developments in study of cephems, oxacephems, penams and sulbactam. Many of the compounds reviewed have potential antibacterial activity
Krisztina M Papp-Wallace et al.
Antimicrobial agents and chemotherapy, 56(11), 5687-5692 (2012-08-22)
Acinetobacter baumannii is an increasingly problematic pathogen in United States hospitals. Antibiotics that can treat A. baumannii are becoming more limited. Little is known about the contributions of penicillin binding proteins (PBPs), the target of β-lactam antibiotics, to β-lactam-sulbactam susceptibility

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service