Skip to Content
Merck
All Photos(1)

Key Documents

Y0000437

Chlorphenamine impurity C

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(3RS)-3-(4-Chlorophenyl)-N-methyl-3-(pyridin-2-yl)propan-1-amine

Sign Into View Organizational & Contract Pricing

Select a Size

15 MG
$319.00

$319.00


Available to ship on07 April 2025Details


Request a Bulk Order

Select a Size

Change View
15 MG
$319.00

About This Item

Empirical Formula (Hill Notation):
C15H17ClN2
CAS Number:
Molecular Weight:
260.76
UNSPSC Code:
41116107
NACRES:
NA.24

$319.00


Available to ship on07 April 2025Details


Request a Bulk Order

grade

pharmaceutical primary standard

API family

chlorpheniramine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Clc1ccc(cc1)C(CCNC)c2ncccc2

InChI

1S/C15H17ClN2/c1-17-11-9-14(15-4-2-3-10-18-15)12-5-7-13(16)8-6-12/h2-8,10,14,17H,9,11H2,1H3

InChI key

UICFCNIVUDNZSW-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Chlorphenamine impurity C EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K J Simons et al.
Journal of pharmaceutical sciences, 73(5), 595-599 (1984-05-01)
The pharmacokinetics and urinary excretion of chlorpheniramine were studied in 11 patients, aged 6-16 years, with allergic rhinitis. In these children, chlorpheniramine had a mean elimination half-life of 13.1 +/- 6.6 h, a mean clearance rate of 7.23 +/- 3.16
B T Hiep et al.
Journal of chromatography. B, Biomedical sciences and applications, 707(1-2), 235-240 (1998-06-05)
The enantiomers of chlorpheniramine and its monodesmethyl metabolite were determined separately in urine by using a coupled achiral-chiral chromatographic system. The two enantiomers of the studied compound and the internal standard were separated from the biological matrix on a cyanopropyl
Skin permeation of chlorpheniramine maleate and detection of demethylated metabolites by high-performance liquid chromatography.
S Zbaida et al.
Journal of pharmaceutical sciences, 77(2), 188-190 (1988-02-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service