Skip to Content
Merck
All Photos(1)

Key Documents

Y0000421

Stavudine for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2′,3′-Didehydro-3′-deoxythymidine, 1-(2,3-Dideoxy-β-D-glycero-pent-2-enofuranosyl)thymine, 1-[(2R,5S)-5-(Hydroxymethyl)-2,5-dihydrofuran-2-yl]-5-methylpyrimidine-2,4-(1H,3H)-dione, 2′,3′-Anhydrothymidine, Stavudine, d4T

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$204.00

$204.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$204.00

About This Item

Empirical Formula (Hill Notation):
C10H12N2O4
CAS Number:
Molecular Weight:
224.21
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$204.00


Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

stavudine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC1=CN([C@@H]2O[C@H](CO)C=C2)C(=O)NC1=O

InChI

1S/C10H12N2O4/c1-6-4-12(10(15)11-9(6)14)8-3-2-7(5-13)16-8/h2-4,7-8,13H,5H2,1H3,(H,11,14,15)/t7-,8+/m0/s1

InChI key

XNKLLVCARDGLGL-JGVFFNPUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Stavudine for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nilza Nascimento Guimarães et al.
Expert opinion on drug safety, 9(5), 771-781 (2010-04-10)
The nucleoside reverse transcriptase inhibitors (NRTIs) are used in antiretroviral therapy worldwide for the treatment of HIV infections. These drugs act by blocking reverse transcriptase enzyme activity, causing pro-viral DNA chain termination. As a consequence, NRTIs could cause genomic instability
R L Murphy
Antiviral therapy, 3 Suppl 4, 69-73 (2000-03-21)
Initial studies of multiple-agent antiretroviral combinations including the thymidine nucleoside analogue reverse transcriptase inhibitor (RTI) stavudine (2',3'-didehydro-2',3'-dideoxythymidine; D4T) have shown potent anti-HIV effects in both treatment-naive and -experienced patients. A number of ongoing randomized comparative trials are assessing stavudine-based multiple
Alain Makinson et al.
Expert opinion on drug safety, 7(3), 283-293 (2008-05-09)
Western randomized trials and prospective cohorts in resource-limited settings have proven virological success with stavudine-based highly active antiretroviral therapy. However, stavudine is no longer recommended in first-line treatments in these two settings due to its intrinsic toxicities and side effects.
M Hurst et al.
Drugs, 58(5), 919-949 (1999-12-14)
Stavudine is a thymidine nucleoside analogue which is phosphorylated intracellularly to an active metabolite, stavudine 5'-triphosphate. This metabolite inhibits HIV replication, either by competing with thymidine 5'-triphosphate for incorporation into viral DNA by reverse transcriptase or by causing premature termination
S A Riddler et al.
Antiviral research, 27(3), 189-203 (1995-06-01)
Stavudine, 2',3'-didehydro-3'-deoxythymidine (D4T), is a potent inhibitor of HIV-1 reverse transcriptase in vitro. In clinical studies, stavudine has excellent oral bioavailability in excess of 80%. The dose-limiting toxicity is peripheral neuropathy, which occurred in 15% of stavudine versus 6% of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service