Skip to Content
Merck
All Photos(1)

Key Documents

Y0000131

Buspirone hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

N-[4-[4-(2-Pyrimidinyl)-1-piperazinyl]butyl]-8-azaspiro[4.5]decane-7,9-dione hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$218.00

$218.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$218.00

About This Item

Empirical Formula (Hill Notation):
C21H31N5O2 · HCl
CAS Number:
Molecular Weight:
421.96
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$218.00


Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

buspirone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl[H].O=C1CC2(CCCC2)CC(=O)N1CCCCN3CCN(CC3)c4ncccn4

InChI

1S/C21H31N5O2.ClH/c27-18-16-21(6-1-2-7-21)17-19(28)26(18)11-4-3-10-24-12-14-25(15-13-24)20-22-8-5-9-23-20;/h5,8-9H,1-4,6-7,10-17H2;1H

InChI key

RICLFGYGYQXUFH-UHFFFAOYSA-N

Gene Information

human ... HTR1A(3350)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Buspirone hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

5-HT1A serotonin receptor agonist; anxiolytic.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Skolnick et al.
Pharmacotherapy, 4(6), 308-314 (1984-11-01)
Buspirone is an azaspirodecanedione that has anxiolytic actions in man and anticonflict actions in experimental animals. In contrast to many commonly used anxiolytic agents, buspirone does not appear to have significant sedative, muscle relaxant or anticonvulsant actions. Consequently, it has
M Kolcsar et al.
Acta physiologica Hungarica, 101(4), 479-487 (2014-12-24)
The role of serotonin in depression and anxiety is still highly controversial. In this experiment the effect of two substances upon anxiety was studied in rats. Forty adult (45-55 weeks old) male Wistar rats were used to study the following
H L Goldberg
Pharmacotherapy, 4(6), 315-324 (1984-11-01)
Buspirone is the first of a new class of anxioselective agents, the azaspirodecanediones. It achieves peak serum concentrations within one hour and has a serum half-life of 2 to 5 hours. Animal studies have suggested antianxiety activity and the absence

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service